Handles
Thermo Scientific™ Shandon™ Stainless-Steel Scalpel Blade Handle, Holds No. 20-25 Blades
Securely hold blades with the Thermo Scientific™ Shandon™ Stainless-Steel Scalpel Blade Handle, constructed of #4 stainless steel.
Thermo Scientific™ Shandon™ Plastic Scalpel Blade Handle, Holds No. 20-25 Blades
Securely hold blades with the autoclavable Thermo Scientific™ Shandon™ Plastic Scalpel Blade Handle, constructed of red plastic #6.
Thermo Scientific™ Shandon™ Plated Scalpel Blade Handle, Holds No. 20-25 Blades
Securely hold blades with the Thermo Scientific™ Shandon™ Plated Scalpel Blade Handle, covered in nickel-plated steel #4.
Fisherbrand™ High Precision #4 Style Scalpel Handle
For use with #22 style blades. Fisherbrand High Precision #4 Style Scalpel Handle is made of stainless steel.
Fisherbrand™ Disposable-Blade Dissecting Knife Handles
Fisherbrand smooth, tapered back end serves as blunt probe
Aspen Surgical™ Bard-Parker™ Surgical Blade Handles
For postmortem work, trimming light plaster casts, and similar forceful cutting
Fisherbrand™ High Precision #3 Style Scalpel Handle Get IT Item
For use with #10 style blades. Fisherbrand High Precision #3 Style Scalpel Handle is made of stainless steel.
Surgical Design Graduated Scalpel Handles
Offers etched graduated markings for convenient measurements
Excelta™ Stainless Steel 5 in. Scalpel Handle
Stainless steel scalpels featuring outside finger serrations
Thermo Scientific™ Shandon™ Nickel-Plated Scalpel Blade Handle, Holds No. 20-25 Blades
Securely hold blades with the Thermo Scientific™ Shandon™ Nickel-Plated Scalpel Blade Handle, constructed of rustless, nickel-plated steel #4. 6.25in. (15.9cm) long.
Thermo Scientific™ Shandon™ 412 Handle and Blades
Cut with the properly hardened, tempered and passivated Thermo Scientific™ Shandon™ 412 Handle and Blades.
Surgical Design Surgical Knife Scalpel Handle, Extra Long
Surgical Design™ Surgical Knife Scalpel Handle (Extra Long) used for sharp and precise dissection. The knife handle comes with patterned surface for enhanced comfort and secure grip that increases precision during use.
Surgical Design Round Scalpel Handles
Provides greater rotational movement with fingertip control
Surgical Design Premier Adjustable Surgical Knife Scalpel Handle
Surgical Design™ Premier Adjustable Surgical Knife Scalpel Handle used for sharp and precise dissection that features a unique multi-angle configuration. The knife handle comes with patterned surface for enhanced comfort and secure non-slip grip.
Surgical Design Royaltek™ Carbon Steel Scalpel Blades
Surgical Design™ Royaltek™ Carbon Steel Scalpel Blades are made from high quality carbon steel and sharpened to perfection. 10 series blades fit 3, 3L, 3 Graduated, 5B, 7, 9 B3, and B3L scalpel handles. 20 series blades fit 4, 4L, 4 Graduated and 6B.
Surgical Design Surgical Knife Scalpel Handle
Surgical Design™ Surgical Knife Scalpel Handle used for sharp and precise dissection. Compatible with 60 and 70 Series blades.
Thermo Scientific™ Shandon™ Plastic Scalpel Blade Handle No. 5, Holds No. 10-15 Blades
Securely hold blades with the Thermo Scientific™ Shandon™ Plastic Scalpel Blade Handle, No. 5, constructed from autoclavable green plastic.
Surgical Design Surgical Knife Set; Set of 8 Scalpel Handles
Surgical Design™ Surgical Knife Set includes the most common and widely used scalpel handles in the field.
Surgical Design Scalpel Blade Remover
Surgical Design™ Scalpel Blade Remover designed for easy, effortless removal of scalple blades from handle
Cincinnati Surgical Stainless Steel Autopsy Handle
Made from high quality British steel
Mopec Scalpel Handle, Bard Parker #8 
Smooth scalpel handle by Mopec for #60 and #70 blades. Durable and reusable.
Mopec Scalpel Handle, Stainless Steel, Smooth #8
Smooth, stainless steel scalpel blade handle by Mopec for #60 and #70 blades. Durable for everyday use.
Mopec Scalpel Handle for Double Edged Blade
Mopec's double edge blade handle is ideal for trimming surgical specimens and is a versatile tool for any laboratory. The double edge blade is securely held in place, yet easy to replace. The weighted pencil-style handle provides a firm, mobile grip.
  spinner