Liquid Samplers
Fisherbrand™ Reusable Polyethylene Dippers
500mL beakers with long handles for removing samples from large tanks.
Bel-Art™ SP Scienceware™ Long-Handled Dippers Available on GSA/VA Contract for Federal Government customers only.
Welded handles. SP Scienceware™ Long-Handled Dippers for easy sampling from tanks, vats, and lakes. These high-density polyethylene plastic dippers will not rust or corrode, provide excellent chemical resistance and float, “just in case.”
Bel-Art™ SP Scienceware™ Water Sample Dipper Available on GSA/VA Contract for Federal Government customers only.
Long reach for otherwise inaccessible spots or taking samples of water, sewage, wastes and effluents from lakes, streams, tanks or ponds
Bel-Art™ SP Scienceware™ Economy Dippers
For convenient sampling from tanks, vats, streams, ponds, lakes
Fisherbrand™ Drum Sampler
For sampling of 55-gallon drums or vessels.
Fisherbrand™ Reusable Polyethylene Ladles
Extra-heavy-duty for sampling and dispensing corrosive liquids.
Fisherbrand™ PTFE Dippers
Totally inert, reusable.
Fisherbrand™ HDPE Liquid Samplers
For industrial and environmental applications.
Corning™ Polypropylene Screw Cap Dipper With Removable Handle, Blue, Sterile
Quality sampling tools for secure sample retrieval and shipment
Bel-Art™ SP Scienceware™ Sample Thief
Hand-controlled polyethylene sampler
Bel-Art™ SP Scienceware™ Ladles
Safely sample and dispense corrosive liquids with these high-density polyethylene plastic ladles.
Bel-Art™ SP Scienceware™ Economy Dipper Cup, Screw-on
Safely gather samples from tanks, vats, streams, rail cars.
DWK Life Sciences Kimble™ Disposable Drum Sampler
Borosilicate glass, open-end drum thieves break easily for quick disposal. Available in 150mL, 25mL and 75mL capacity.
DWK Life Sciences Wheaton™ Stainless-Steel Bomb Sampler
Collect liquid or light viscous samples from tanks, ponds, lagoons and wells
Bel-Art™ SP Scienceware™ Liquid Sampler
A 50mL (1.65oz) liquid sampler made for immersion into a can, bottle, tank, vat, etc.
Fisherbrand™ PTFE Sampler/Ladle
Made from pure PTFE.
Bel-Art™ SP Scienceware™ Safety Acid/Caustic Sampler
Well-suited for sodium hydroxide, potassium hydroxide, hydrochloric acid, acetic acid, formaldehyde, sulfuric acid, etc.
Fisherbrand™ Reusable Polyethylene Dippers Products for Science Education
500 mL beakers with long handles for removing samples from large tanks.
Fisherbrand™ PTFE Dippers Products for Science Education
Totally inert, reusable.
Fisherbrand™ Drum Sampler Products for Science Education
For sampling of 55-gallon drums or vessels.
Fisherbrand™ Reusable Polyethylene Ladles Products for Science Education
Extra-heavy-duty for sampling and dispensing corrosive liquids.
  spinner