Gas Needle Valves
DWK Life Sciences Kimble™ Kontes™ AIRLESS-WARE™ Pressure Release Valve
Designed for applications that require evacuated apparatus to be filled with an inert gas to one atmosphere. Fits 3/8 in. tubing.
  spinner