Tweezers and Forceps
Fisherbrand™ General Purpose Tweezers, 13cm Long
For precision handling without metal contact.
Fisherbrand™ General Purpose Tweezers, 12.5cm Long
For precision handling without metal contact.
Thermo Scientific™ Nalgene™ Stainless-Steel Forceps Available on GSA/VA Contract for Federal Government customers only.
Nalgene Stainless-Steel Forceps are available in bent or straight-tip styles. Bent-tip style is designed for easy removal and manipulation of filter membranes.
Fisherbrand™ General Purpose Tweezers, 12cm Long
For precision handling without metal contact.
Thermo Scientific™ Nalgene™ Polypropylene Scissor-Type Forceps Available on GSA/VA Contract for Federal Government customers only.
Avoid corrosion from acid exposure with Thermo Scientific™ Nalgene™ Polypropylene Scissor-Type Forceps, which are chemically compatible with most lab reagents.
Fisherbrand™ Dissecting Forceps
Bone-cutting stainless steel
Bel-Art™ SP Scienceware™ Polypropylene Forceps Available on GSA/VA Contract for Federal Government customers only.
Serrated tips to pick up sample with a firm grip
Fisherbrand™ Straight Strong Tip General Purpose Forceps
Popular general-purpose tweezers for use without magnification
Bel-Art™ SP Scienceware™ PTFE-Coated Steel Forceps
Stainless-steel forceps with Fluo-Kem™ PTFE FEP coated tips
Fisherbrand™ Offset Flat-Tip Forceps
45° offset is ideal for picking up flat objects off a flat surface
Fisherbrand™ Curved Tapered Flat-Tip Forcep
Curved tip for added visibility
Fisherbrand™ High Precision Tissue Forceps with Serrated Tips
Used for holding tissues during procedures. Fisherbrand High Precision Tissue Forceps with Serrated Tips are made of stainless steel.
Fisherbrand™ Fine Precision General-Purpose Assembly Forceps
Includes miniature, 3.5 in. (90mm), model
Fisherbrand™ Straight Flat Tip Forceps
Designed for handling labels or SMD components
Fisherbrand™ General-Purpose PTFE Forceps
For use up to 260°C (500°F)
Fisherbrand™ General-Purpose Curved Forceps
Heavy-duty type for general lab work
Fisherbrand™ General-Purpose Extra-Long Forceps
Remove specimens from deep bottles or jars with long forceps
EMD Millipore Filter Forceps
Handle membrane filters without damage
Fisherbrand™ Curved Very Fine Precision Tip Forceps
Allows maximum visibility under magnified applications
Fisherbrand™ High Precision Straight Very Fine Point Tweezers/Forceps
Designed for work under magnification. Fisherbrand High Precision Straight Very Fine Point Tweezers/Forceps is made of anti-magnetic stainless steel.
Fisherbrand™ Filter Forceps
Easy, effortless handling of membranes and glass fiber filters
Fisherbrand™ High Precision Straight Broad Strong Point Tweezers/Forceps
Ideal for use in general applications. Fisherbrand High Precision Straight Broad Strong Point Tweezers/Forceps is made of anti-magnetic stainless steel.
Fisherbrand™ Straight Broad Strong Tip General Application Forceps
Feature serrated grips for superior control. Fisherbrand™ Straight Broad Strong Tip General Application Forceps are easy to use and available in 6 or 12 inch varieties.
Fisherbrand™ High Precision 45° Curved Tapered Very Fine Point Tweezers/Forceps
Allows maximum visibility under magnified applications. Fisherbrand High Precision 45° Curved Tapered Very Fine Point Tweezers/Forceps is made of anti-magnetic stainless steel.
Fisherbrand™ Straight Very Fine Point Reverse Action Forceps
Closed at rest and require finger pressure to open
  spinner