Capillary Electrophoresis System Accessories
Applied Biosystems™ Cathode Buffer Container For 3500 Series Genetic Analyzers Non-distribution product as customer accommodation.
The CBC is made in a ready-to-use, disposable container with a radio frequency identification (RFID) tag incorporated into the label.
Applied Biosystems™ GeneScan™ 400HD ROX™ dye Size Standard Non-distribution product as customer accommodation.
The GeneScan™ 400HD ROX™ Size Standard is a ROX™ dye-labeled size standard for the reproducible sizing of fragment analysis data. The standard contains 21 ROX™ dye-labeled DNA fragments.
Applied Biosystems™ GeneScan™ Installation Standard DS-33 , [6FAM™, VIC™, NED™, PET™ dyes] Non-distribution product as customer accommodation.
Using 6FAM™, VIC®, NED™ and PET® dyes contain pooled PCR products amplified from microsatellite loci present in CEPH individual 1347.
Applied Biosystems™ SeqStudio™ Cartridge Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
The Applied Biosystems SeqStudio Cartridge is an all-in-one and easy-to-use reagent cartridge that includes capillaries, universal polymer, buffer, and pump required to run on the SeqStudio Genetic Analyzer System.
Applied Biosystems™ SeqStudio™ Cathode Buffer Container Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
The Applied Biosystems SeqStudio Cathode Buffer Container (CBC) contains running buffer to support Sanger sequencing and fragment analysis applications on the SeqStudio Genetic Analyzer System.
Applied Biosystems™ 4-Capillary Array For 3130/3100-Avant™ Genetic Analyzer Non-distribution product as customer accommodation.
Use for fragment analysis and sequencing. Usage per array is 100 runs.
Applied Biosystems™ Septa Cathode Buffer Container For SeqStudio™ Genetic Analyzer Non-distribution product as customer accommodation.
This Buffer Container is for the SeqStudio™ Genetic Analyzer
Applied Biosystems™ 96-well Septa for SeqStudio™ Genetic Analyzer Non-distribution product as customer accommodation.
These are 96-well plate septa for use on the SeqStudio Genetic Analyzer.
Applied Biosystems™ 8-strip Septa For SeqStudio™ Genetic Analyzer Non-distribution product as customer accommodation.
A radio-frequency identification tag is built into the frame for tracking critical run information, which is then reported on the software dashboard.
Applied Biosystems™ SeqStudio™ Integrated Capillary Protector Non-distribution product as customer accommodation.
The Applied Biosystems SeqStudio Integrated Capillary Protector (ICP) is used on the SeqStudio Cartridge. Gel-filled and is for one-time-use only.
Applied Biosystems™ GeneScan™ 120 LIZ™ Dye Size Standard Non-distribution product as customer accommodation.
GeneScan 120 LIZ Size Standard is designed for sizing DNA fragment in the 15-120 nucleotides range and provides nine single-stranded labeled fragment.
  spinner