Capillary Electrophoresis Systems
Applied Biosystems™ SeqStudio™ Genetic Analyzer System with SmartStart and Sanger Sequencing Kit Non-distribution product as customer accommodation.
The Applied Biosystems SeqStudio Genetic Analyzer is a 4-capillary, fluorescence-based capillary electrophoresis system designed to provide users with an integrated experience that is easy, efficient, and versatile for research in genetic analysis. This simple, fast workflow enables completion of Sanger sequencing, from sample to answer, in less than a workday.
Applied Biosystems™ USB Sequenase Reaction Buffer Non-distribution product as customer accommodation.
The USB Sequenase Reaction Buffer (5X) is a component of the Sequenase Quick Denature Plasmid DNA Sequencing Kit. This buffer can also be purchased separately.
Applied Biosystems™ SeqStudio™ Starter Kit for SeqStudio™ Genetic Analyzer Available on GSA/VA Contract for Federal Government customers only.
This is the Starter Kit that is recommended for purchase with the SeqStudio
Applied Biosystems™ Centri-Sep™ 96-well Plate Non-distribution product as customer accommodation.
Centri-Sep™ 96's hydrated matrix enables effective and reliable high-throughput removal of excess BigDye™terminators from DNA sequencing reactions, giving you clean sequencing reads with uniform signal intensity starting with the first base.
Applied Biosystems™ SeqStudio™ Laptop Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
This is the optional LapTop Computer (purchases separately) for the SeqStudio
Applied Biosystems™ SeqStudio™ Genetic Analyzer System with SmartStart Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
The Applied Biosystems SeqStudio Genetic Analyzer is a 4-capillary, fluorescence-based capillary electrophoresis system designed to provide users with an integrated experience that is easy, efficient, and versatile for research in genetic analysis.
  spinner