Flash Point Testers
Koehler™ Rapid Flash Closed-Cup Tester
Verifies the flash point or the sustained burning qualities of small samples in the range of -30°C to +300°C.
Koehler™ Tag Open Tester
Provides excellent reproducibility for accurate test results.
Koehler™ Cleveland Open-Cup Flash Tester
For flash and fire points of all petroleum products, except fuel oils and those having an open cup flash below 79°C (175°F)
Koehler™ Pensky-Martens Closed Tester
For flash point determinations of fuels, lubricating oils, liquids containing suspended solids and liquids that tend to form a surface film during testing
Koehler™ Tag Closed Tester
Provides the simple, accurate control of heating rate necessary for ASTM D 56 in determining flash point of liquids below 93°C (200°F).
  spinner