Fluorometers
Invitrogen™ Qubit™ Assay Tubes Non-distribution product as customer accommodation.
For use with the Qubit 2.0 Fluorometer
Thermo Scientific™ NanoDrop™ 3300 Fluorospectrometer Available on GSA/VA Contract for Federal Government customers only.
Choose this versatile, cost-effective fluorospectrometer for broad spectrum fluorescence analysis.
Promega™ Replacement AC Adapter for QuantiFluor™-ST
For use with the QuantiFluor-ST Single-Tube Fluorometer.
Promega™ Solid Standard for QuantiFluor™-ST Fluorometer
Designed to verify the stability of the fluorometer by checking drift of the calibration.
Promega™ Quantus™ Fluorometer
Dual-channel fluorometer for personal quantitation workflow. Designed to provide highly sensitive fluorescent detection when quantifying nucleic acids, the compact instrument is simple to operate. The Quantus Fluorometer is optimized with preprogrammed settings for Promega QuantiFluor™ Dye Systems to quantitate nucleic acids and offers the flexibility to create customized methods and quantitation settings for other fluorescent dyes.
Promega™ QuantiFluor™-ST and QuantiFluor-P Single-Tube Fluorometers: PCR Tube Adapter
PCR Tube Adapter for QuantiFluor single-tube fluorometers
  spinner