Bulk Tray Dryers
Labconco™ Tube Holders for Fast-Freeze™ and Lyph-Lock™ Flasks
Holds tubes in upright position for freeze drying.
Labconco™ FreeZone™ Bulk Tray
For FreeZone Bulk Tray Dryers and/or Stoppering Tray Dryers
Labconco™ FreeZone™ Small Tray Dryers
Designed for lyophilizing moderate to large bulk sample loads. Labconco™ FreeZone™ Small Tray Dryers offer options for a built-in 6-port manifold and isolation valve. They include three heated shelves and temperature probes for monitoring sample temperature.
Labconco™ FreeZone™ Bulk Tray Dryers, 6-Port Manifold & Isolation Valve, 230V Models
Designed for lyophilizing moderate to large bulk sample loads. Labconco™ FreeZone™ Bulk Tray Dryers, 6-Port Manifold & Isolation Valve, 230V Models offer options for built-in 6-port manifold and isolation valve. They include three heated shelves and temperature probes for monitoring sample temperature.
Labconco™ FreeZone™ Stoppering Tray Dryers, 230V Models
Provides precise control and program flexibility for consistent performance. Labconco™ FreeZone™ Stoppering Tray Dryers, 230V Models are designed for lyophilizing moderate to large loads and stoppering vials or serum bottles under vacuum. They offer options for built-in 6-port manifold and isolation valve.
Labconco™ FreeZone™ Stoppering or Bulk Tray Dryer Accessories
Accessories for FreeZone Stoppering and Bulk Tray Dryers.
Labconco™ FreeZone™ Bulk Tray Dryers, 6-Port Manifold, 230V Models
Designed for lyophilizing moderate to large bulk sample loads. Labconco™ FreeZone™ Bulk Tray Dryers, 6-Port Manifold, 230V Models offer options for built-in 6-port manifold and isolation valve. They include three heated shelves and temperature probes for monitoring sample temperature.
Labconco™ FreeZone™ Stoppering Tray Dryers, US Models
FreeZoneStoppering Tray Dryers provide precise control and program flexibility for consistent performance and are designed for lyophilizing moderate to large loads and stoppering vials or serum bottles under vacuum. They offer options for a built-in 6-port manifold and isolation valve.
Labconco™ Small Tray: FreeZone™ Stoppering or Bulk Tray Dryer Accessory
Small Tray designed for use with the FreeZone™ Tray Dryer.
Labconco™ FreeZone™ Bulk Tray Dryers, Isolation Valve, 230V Models
Designed for lyophilizing moderate to large bulk sample loads. Labconco™ FreeZone™ Bulk Tray Dryers, Isolation Valve, 230V Models offer options for built-in 6-port manifold and isolation valve. They include three heated shelves and temperature probes for monitoring sample temperature.
Labconco™ FreeZone™ Bulk Tray Dryers, 230V Models
Designed for lyophilizing moderate to large bulk sample loads. Labconco™ FreeZone™ Bulk Tray Dryers, 230V Models offer options for built-in 6-port manifold and isolation valve. They include three heated shelves and temperature probes for monitoring sample temperature.
Labconco™ FreeZone™ Stoppering Tray Dryers, 6-Port Manifold, 230V Models
Provides precise control and program flexibility for consistent performance. Labconco™ FreeZone™ Stoppering Tray Dryers, 6-Port Manifold, 230V Models are designed for lyophilizing moderate to large loads and stoppering vials or serum bottles under vacuum. They offer options for built-in 6-port manifold and isolation valve.
Labconco™ 6-Port Manifold and Isolation Valve
For use with FreeZone Tray Dryers and Bulk Tray Dryers
Labconco™ FreeZone™ Stoppering Tray Dryers, 6-Port Manifold and Isolation Valve, 230V Models
Provides precise control and program flexibility for consistent performance. Labconco™ FreeZone™ Stoppering Tray Dryers, 6-Port Manifold and Isolation Valve, 230V Models are designed for lyophilizing moderate to large loads and stoppering vials or serum bottles under vacuum. They offer options for built-in 6-port manifold and isolation valve.
Labconco™ FreeZone™ Stoppering Tray Dryers, Isolation Valve, 230V Models
Provides precise control and program flexibility for consistent performance. Labconco™ FreeZone™ Stoppering Tray Dryers, Isolation Valve, 230V Models are designed for lyophilizing moderate to large loads and stoppering vials or serum bottles under vacuum. They offer options for built-in 6-port manifold and isolation valve.
Labconco™ FreeZone™ Bulk Tray Dryers, US Models
Designed for lyophilizing moderate to large bulk sample loads. Labconco™ FreeZone™ Bulk Tray Dryers, US Models offer options for built-in 6-port manifold and isolation valve. They include three heated shelves and temperature probes for monitoring sample temperature.
Labconco™ FreeZone™ Shelf For Bulk Tray Dryer
Additional shelf for Bulk Tray Dryers only
Labconco™ FreeZone™ Bulk Tray Dryers
For use with 6, 12, or 18L FreeZone Freeze-Dry Systems
  spinner