Hydrometers
Fisherbrand™ Traceable™ Digital Hygrometer/Thermometer
Hang on the wall or use included plastic stand; housed in rugged ABS case
Fisherbrand™ Specific Gravity Hydrometers Available on GSA/VA Contract for Federal Government customers only.
Double scales can be read without turning the hydrometer.
Fisherbrand™ Set of Eight Precision Specific Gravity Hydrometers Available on GSA/VA Contract for Federal Government customers only.
Specific Gravity Hydrometers are permanently marked with individual serial numbers.
Fisherbrand™ Specific Gravity/Baume Hydrometers Available on GSA/VA Contract for Federal Government customers only.
Fisherbrand™ Specific Gravity/Baume Hydrometers are calibrated according to ASTM™ E 126 specifications.
Fisherbrand™ Baume Hydrometers Available on GSA/VA Contract for Federal Government customers only.
Baume “heavy” scale for working with liquids heavier than water.
H-B Instrument™ Durac™ API ASTM Hydrometers with Individual Calibration Report; Traceable to NIST
Measure the relative density of oil and petroleum products in ranges from -1° to 101° API with these hydrometers. H-B Instrument Durac™ API ASTM Hydrometers conform to American Petroleum Institute (API) and the American Society of Testing Materials (ASTM) E100 standards. Traceable to NIST, with Individual Calibration Report.
H-B Instrument™ Durac™ Baume Plastic Plain Form Hydrometers
Safety and quality protection in one instrument!
H-B Instrument™ DURAC™ Safety Precision Combined Form Brix Sugar Hydrometers
These combined-form hydrometers are used to measure % sucrose by weight in various ranges from 0° to 101° Brix, and include a convenient, built-in thermometer with a temperature scale of 0/50°C
Fisherbrand™ Soil Analysis ASTM Hydrometers
These hydrometers are used to determine the specific gravity of soil in the range of 0.995 to 1.038 or -5 to 60g/L (using a weight by volume scale).
Fisherbrand™ Midget Urinometer Available on GSA/VA Contract for Federal Government customers only.
Urinometer accuracy guaranteed to ±0.002.
H-B Instrument™ Durac™ Salt Brine Plastic Plain Form Hydrometers
Lead free and shatterproof for ultimate safety
Fisherbrand™ Salimeters Available on GSA/VA Contract for Federal Government customers only.
Fisherbrand™ Salimeters are used to measure pickling and meat-packing brines.
Fisherbrand™ Alcohol Hydrometers Available on GSA/VA Contract for Federal Government customers only.
Used by distillers, law enforcement and taxing agencies to measure the alcoholic content of beverages.
H-B Instrument™ Durac™ ASTM Precision Specific Gravity Hydrometers with Individual Calibration Report: for Heavy Liquids
Available in a variety of individually calibrated models measuring specific gravity of heavy liquids in various ranges from 0.650 to 1.500
Fisherbrand™ Replacement Hydrometer for Squibb Urinometer Available on GSA/VA Contract for Federal Government customers only.
For use with Fisherbrand™ Squibb Urinometer (cat. no. 22274233).
H-B Instrument™ Durac™ Specific Gravity/Relative Density (g/cm&superscript_3;) Plain Form Hydrometers: for Liquids Heavier Than Water
Available in a variety of models measuring specific gravity/relative density (g/cm&superscript_3;) of liquids heavier than water in various ranges.
Fisherbrand™ Plain Form Petroleum Hydrometers Available on GSA/VA Contract for Federal Government customers only.
API Plain Form Petroleum Hydrometers are available in regular and ASTM™ recommended forms.
H-B Instrument™ Durac™ API ASTM Hydrometers
Measures relative densities (specific gravity) of oil and petroleum products in accordance with American Petroleum Institute (API) and American Society of Testing Materials (ASTM) E100 standards
DWK Life Sciences Kimble Chase™ Short Form High Precision Specific Gravity Hydrometers
Available in a variety of graduation ranges and overall lengths
DWK Life Sciences Kimble Chase™ Baume/SG Dual Scale Hydrometers
Available in a variety of graduation ranges and overall lengths
Fisherbrand™ ASTM™ Specific Gravity Hydrometers for Liquids Lighter than Water Available on GSA/VA Contract for Federal Government customers only.
Accuracy of hydrometer meets or exceeds ASTM™ E 100 specifications.
Fisherbrand™ ASTM™ Hydrometers, Plain Form Available on GSA/VA Contract for Federal Government customers only.
API hydrometers are available in regular and ASTM™ recommended forms.
DWK Life Sciences Kimble Chase™ Precision Specific Gravity Hydrometers
Available in a variety of graduation ranges and overall lengths
DWK Life Sciences Kimble Chase™ Ethanol Hydrometer
With dual graduations in 0 to 100% and 0 to 200 proof scales
  spinner