Osmometers
Precision Systems Osmette II™ and Osmette III™ Osmometers
The simplicity and dependability of Osmette™ osmometers in a small, lightweight size. Osmette II and Osmette III Osmometers are ideal for one-step freezing-point measurement of osmolality.
Advanced Instruments Micro-Sample Test Kit
The Advanced Instruments Micro-Sample Test Kit ensures accurate and precise results with the Advanced Instruments Osmo1 and 3320 Single-Sample Micro-Osmometers.
Clinitrol™ 290 Reference Solution; Advanced Instruments Available on GSA/VA Contract for Federal Government customers only.
Critical normal human serum midpoint of 290mOsm
Advanced Instruments Osmometer Disposables Available on GSA/VA Contract for Federal Government customers only.
Disposables for use with Advanced osmometers
Precision Systems Printer Paper for 5010 Osmette III™ Fully Automatic 10μL Osmometer
Accessory for 5010 Osmette III fully automatic 10μL Osmometer.
Advanced Instruments Osmometer Calibration Standards Available on GSA/VA Contract for Federal Government customers only.
Calibration standards for use with Advanced osmometers
Advanced Instruments Probe Wiper Rings Available on GSA/VA Contract for Federal Government customers only.
Probe Wiper Rings for Advanced Multi-Sample Osmometer, Model 2020
Precision Systems Osmette™ Osmometer Disposable Sample Tubes
Sample tubes for use with the Osmette Osmometer.
Advanced Instruments Protinol™ Serum Osmometer Controls Available on GSA/VA Contract for Federal Government customers only.
Quality control check for the clinical laboratory for osmometers from any manufacturer
Advanced Instruments Advanced™ Ease-Eject™ Sampler Available on GSA/VA Contract for Federal Government customers only.
For use with Model 3300 and Model 3320 Micro-Osmometers Only
Advanced Instruments Osmometer General Accessories and Supplies Available on GSA/VA Contract for Federal Government customers only.
Accessories for use with Advanced Instuments Osmometers.
Advanced Instruments Osmolality Linearity Set Available on GSA/VA Contract for Federal Government customers only.
For monitoring performance of any osmometer in the range 0-2000 mOsm/kg
Advanced Instruments Probe Wiper Discs
Supplies for A2O™ Advanced Automated Osmometer
Advanced Instruments Osmometer System Fluid
For A2O™ Advanced™ Automated Osmometer
Precision Systems Bath Fluid for Osmette™ Osmometers
For use with Osmette A Osmometer, Osmette XL Osmometer.
Advanced Instruments Osmometer Standards
For in vitro diagonostic use
Advanced Instruments Sampler Calibration Key for Advanced™ 3320 Micro Osmometer
Replacement calibration key for Advanced 3320 Micro Osmometer 20μL sampler
Advanced Instruments Osmometer Printers and Accessories
Printer paper and cartridge for use with Advanced Osmometer Printer.
Precision Systems Osmette™ Multi-Osmette Osmometer
Automatic digital osmometers measure osmolality in mOsm/kg H2O. Osmette Multi-Osmette Osmometer uses the freezing point depression method to provide a quick, accurate indication of total concentration. Instruments measure freezing points of solutions to 0.001°C, then directly translate them to measure of concentration.
Precision Systems Osmette™ XL Osmometer
Automatic digital osmometers measure osmolality in mOsm/kg H2O. Precision Systems Osmette XL Osmometer uses the freezing point depression method to provide a quick, accurate indication of total concentration. Instruments measure freezing points of solutions to 0.001°C, then directly translate them to measure of concentration.
Precision Systems Osmette™ A Osmometer
Automatic digital osmometers measure osmolality in mOsm/kg H2O. Osmette A Osmometer uses the freezing point depression method to provide a quick, accurate indication of total concentration. Instruments measure freezing points of solutions to 0.001°C, then directly translate them to measure of concentration.
Advanced Instruments OsmoPRO™ Multi-Sample Micro-Osmometer
Simple and effiientc freezing point osmometer. The Advanced Instruments OsmoPRO Multi-Sample Micro-Osmometer is ideal for mid- to high-volume laboratories that prefer to automate osmolality batch testing while improving laboratory effciency, throughput, a
Advanced Instruments Model 3250 Single-Sample Osmometer Available on GSA/VA Contract for Federal Government customers only.
Provides fast, accurate test results using 200–250μL
Precision Systems Osmette™ Micro-Osmette Osmometer
Automatic digital osmometers measure osmolality in mOsm/kg H2O. Osmette Micro-Osmette Osmometer uses the freezing point depression method to provide a quick, accurate indication of total concentration. Instruments measure freezing points of solutions to 0.001°C, then directly translate them to measure of concentration.
Advanced Instruments Osmo1 Single-Sample Micro-Osmometer
The Advanced Instruments Osmo1 Single-Sample Micro-Osmometer offers direct draw of small sample volumes, ease-of-use, accurate and precise results, and the security and efficiency of electronic data management.
  spinner