Photometers and Light Meters
Fisherbrand™ Traceable™ Dual-Range Light Meter
Measures both lux and foot-candles at a rapid sampling rate of 0.4 seconds. Display update rate is adjustable to 1 or 2 seconds.
Fisherbrand™ Enviro-Meter™
Functions as light meter, anemometer, humidity meter, and thermometer—all in one unit
Fisherbrand™ Traceable™ UV Light Meter
Easy-to-use meter with a wide-band UV wavelength of 320 to 390nm.
Fisherbrand™ Traceable™ Dual-Display Light Meter
Large LCD shows separate readings for lux and foot-candles in 1.375 in. H digits, plus selected source (daylight, tungsten, fluorescent or mercury).
Oakton™ Digi-Sense™ Precalibrated Data Logging Light Meter
Ensure lighting standards and increase system efficiencies with precision measurement accuracy
UVP UVX Radiometer: UVX-36 Sensor
Digital UVX Radiometer connects to one of the three interchangeable sensors (sold separately) to measure 254nm, 365nm or 302nm UV wavelengths
UVP UVX Radiometer
Digital UVX Radiometer connects to one of the three interchangeable sensors (sold separately) to measure 254nm, 365nm or 302nm UV wavelengths
UVP UVX Radiometer: UVX-25 Sensor
Digital UVX Radiometer connects to one of the three interchangeable sensors (sold separately) to measure 254nm, 365nm or 302nm UV wavelengths
UVP UVX Radiometer: UVX-31 Sensor
Digital UVX Radiometer connects to one of the three interchangeable sensors (sold separately) to measure 254nm, 365nm or 302nm UV wavelengths
UVP J-Series Photovoltaic UV Intensity Meters: Model J-225
Photovoltaic J-Series UV meters for taking accurate and repeatable readings of UV light sources in either the 365nm or 254nm UV range
YSI™ Portable Photometers
Portable photometers, allow easy readings directly in the field for 100+ parameters
UVP UVX Radiometer: Attenuator
Digital UVX Radiometer connects to one of the three interchangeable sensors (sold separately) to measure 254nm, 365nm or 302nm UV wavelengths
YSI™ EcoSense 9300 Benchtop Photometer
Portable photometers allow to easily take readings directly in the field for 100+ parameters.
UVP J-Series Photovoltaic UV Intensity Meters: Filter Contrast
Replacement part for Photovoltaic J-Series UV meters
YSI™ USB Power Supply Unit for Photometry
For use with 9500 photometer.
YSI™ Starter Nickel Reagent Test Kits for EcoSense 9300/9500 Photometer
Nickel test kit for use with for EcoSense 9300 or 9500 Photometers. Eliminate sample preservation by taking your portable, waterproof instruments to the field to perform the analysis on-site. All reagents are safe and easy to use.
UVP J-Series Photovoltaic UV Intensity Meters: Model J-221
Photovoltaic J-Series UV meters for taking accurate and repeatable readings of UV light sources in either the 365nm or 254nm UV range
Fisherbrand™ Pocket Digital Light Meter Products for Science Education
Compact meter reads from 0 to 1999 foot-candles with resolution from 0.1 to 1.
Fisherbrand™ Traceable™ Dual Display Light Meter Products for Science Education
Zero reference settings for four light sources: tungsten, fluorescent, daylight and mercury. Fisherbrand Traceable Light Meter has two displays with easy-to-read, jumbo-sized 1.38 in. high (3.5cm) digits show lux or foot-candles and the selected light source (daylight, tungsten, fluorescent or mercury).
Light and Moisture Meter Products for Science Education
Instantly measures soil moisture and light conditions — no batteries required.
  spinner