Nuclear Magnetic Resonance Spectrometers
Bel-Art™ SP Scienceware™ Wilmad Luer Lock Syringe Needles Available on GSA/VA Contract for Federal Government customers only.
Designed for easy sample loading with Luer Lock syringes, these needles are available in stainless steel or PTFE. Bel-Art SP Scienceware™ Wilmad Luer Lock Syringe Needles are ideal for puncturing septum in air-sensitive samples.
Bel-Art™ SP Scienceware™ Wilmad™ 150mL Suprasil Offset Nitrogen Dewar Flask for Bruker Available on GSA/VA Contract for Federal Government customers only.
Designed for cavities that have limited space between the reservoir and the drive shaft for iris. Bel-Art SP Scienceware™ Wilmad™ 150mL Suprasil Offset Nitrogen Dewar Flasks provide a convenient alternative to VT systems for studies at 77 K, and are silvered around the reservoir.
Bel-Art™ SP Scienceware™ Wilmad Rotor and Caps for Bruker MAS Probe Available on GSA/VA Contract for Federal Government customers only.
Eliminate the need to solvate with MAS rotors manufactured from the highest purity Zirconia. Bel-Art SP Scienceware™ Wilmad MAS Rotors provide the ultimate solution for analysis of solid samples. Available for most present solid state NMR spectrometers.
Bel-Art™ SP Scienceware™ Wilmad™ Bottom Plug for 3.2mm MAS Rotor Available on GSA/VA Contract for Federal Government customers only.
Designed for use with MAS Rotors. Bel-Art SP Scienceware™ Wilmad™ Bottom Plug for 3.2mm MAS Rotor features a Vespal composition and works with applications ranging in temperature from -30° to 70°:C.
Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ 3 mm Kel-F Positioning Rod Microcell Small Volume NMR Insert Available on GSA/VA Contract for Federal Government customers only.
Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ 3 mm Kel-F Positioning Rod Microcell Small Volume NMR Insert is ideal for experiments that need to center small volume samples in the Rf coil such as magnetic susceptibility measurement.
Bel-Art™ SP Scienceware™ Wilmad™ Pyrex™ MAS Rotor Inserts Available on GSA/VA Contract for Federal Government customers only.
Designed for air-sensitive samples and semi-solid samples such as gels or highly viscous liquids. Bel-Art SP Scienceware™ Wilmad™ Pyrex™ MAS Rotor Inserts allow samples to be sealed into an NMR rotor by heat-seal or epoxy.
Bel-Art™ SP Scienceware™ WIlmad™ OmniFit™ Valve NMR System Available on GSA/VA Contract for Federal Government customers only.
Designed for easy injection of chemicals through a gas-tight syringe for air sensitive samples without using a glove box. Bel-Art SP Scienceware™ WIlmad™ OmniFit™ Valve NMR System is comprised of a 5mm end cap, 2-way valve, septum with cap, barrel coupler and a vacuum connector.
Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ 4 mm Finger for Varian/Bruker Available on GSA/VA Contract for Federal Government customers only.
Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ 4 mm Finger for Varian/Bruker is manufactured to precision standards that exceed spectrometer products in terms of dimension tolerance, reliability and life span.
Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ 3 mm Varian Spinner Turbine Available on GSA/VA Contract for Federal Government customers only.
Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ 3 mm Varian Spinner Turbine is manufactured to precision standards that exceed spectrometer products in terms of dimension tolerance, reliability and life span.
Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ 5 mm Bruker Spinner Turbine Available on GSA/VA Contract for Federal Government customers only.
Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ 5 mm Bruker Spinner Turbine is manufactured to precision standards that exceed spectrometer products in terms of dimension tolerance, reliability and life span.
Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ 3 mm Finger for Varian/Bruker Available on GSA/VA Contract for Federal Government customers only.
Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ 3 mm Finger for Varian/Bruker is manufactured to precision standards that exceed spectrometer products in terms of dimension tolerance, reliability and life span.
Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ O-Ring for Spinner Available on GSA/VA Contract for Federal Government customers only.
Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ O-Ring for Spinner is manufactured to precision standards that exceed spectrometer products in terms of dimension tolerance, reliability and life span.
Bel-Art™ SP Scienceware™ Wilmad™ 4.0mm Solid State NMR Rotor Cap Remover Available on GSA/VA Contract for Federal Government customers only.
Eliminate possible damage of the cap and rotor with this zero turn cap remover. Bel-Art SP Scienceware™ Wilmad™ 4.0mm Solid State NMR Rotor Cap Remover simplifies removal of end caps and base plugs in solid state NMR.
Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ 4 mm Varian Spinner Turbine Available on GSA/VA Contract for Federal Government customers only.
Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ 4 mm Varian Spinner Turbine is manufactured to precision standards that exceed spectrometer products in terms of dimension tolerance, reliability and life span.
Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ Replacement O-Ring for Spinner Available on GSA/VA Contract for Federal Government customers only.
Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ Replacement O-Ring for Spinner is manufactured to precision standards that exceed spectrometer products in terms of dimension tolerance, reliability and life span.
Bel-Art™ SP Scienceware™ Wilmad™ NMR Tube pH Electrode Available on GSA/VA Contract for Federal Government customers only.
Measure pH range from 0 to 14 inside an NMR sample with this electrode. Bel-Art SP Scienceware™ Wilmad™ NMR Tube pH Electrodes are compatible with 5mm Thin Walled NMR tubes.
  spinner