Spectrometer Accessories
Thermo Scientific™ Crystal Plates and Spare Parts for the Thermo Scientific™ iD7 ATR Accessory
Choose the crystal plate to meet specific application and budget requirements
Fisherbrand™ Hellmanex™ III Liquid Cleaning Concentrate
Provides effective cleaning of glass and quartz cells and other sensitive optical components
SPEX™ SamplePrep Disposable XRF X-Cell™ Sample Cups Window Film Available on GSA/VA Contract for Federal Government customers only.
Strong and durable window film. SPEX SamplePrep Disposable XRF X-Cell Sample Cups Window Film covers the cell and serves as the sample window, allowing uniform transmission of X-rays.
SPEX™ SamplePrep Prep-Aid™ Grinding Aids Available on GSA/VA Contract for Federal Government customers only.
Ideal for samples which crumble or ablate after pressing.
SPEX™ SamplePrep Disposable XRF Spec-Cap™ Sample Cups Available on GSA/VA Contract for Federal Government customers only.
Aluminum cups aid in the production of pressed-powder sample discs for XRF.
Eppendorf™ Vis Cuvettes Available on GSA/VA Contract for Federal Government customers only.
Disposable cuvettes made of clear plastic with a light transmission of 300 nm to 900 nm. The Vis cuvettes are the perfect tool for applications outside of the UV range, for example, colorimetric protein assays (Bradford, Lowry, etc.), determining of the optical density of bacterial cultures (OD 600 methods), kinetic, and fluorescence measurements.
SPEX™ SamplePrep Disposable XRF X-Cell™ Sample Cups Available on GSA/VA Contract for Federal Government customers only.
Permit fast XRF analysis of liquid samples.
Alfa Aesar™ Disposable IR Cards, 5x10cm, KBr window, 9.5mm aperture
Provide a conveniently mounted, non-absorbing substrate for qualitative infrared analysis.
Alfa Aesar™ Disposable IR Cards, 5x10cm, NaCl window, 15mm aperture
Provide a conveniently mounted, non-absorbing substrate for qualitative infrared analysis.
Alfa Aesar™ Disposable IR Cards, 5x10cm, NaCl window, 9.5mm aperture
Provide a conveniently mounted, non-absorbing substrate for qualitative infrared analysis.
Thermo Scientific™ Golden Gate ATR for Nicolet™ iS5 FT-IR Spectrometer
This versatile infrared sampling accessory is fast, sensitive, and robust
Alfa Aesar™ Disposable IR Cards, 5x10cm, KCl window, 19mm aperture
Provide a conveniently mounted, non-absorbing substrate for qualitative infrared analysis.
Alfa Aesar™ Disposable IR Cards, 5x10cm, KCl window, 9.5mm aperture
Provide a conveniently mounted, non-absorbing substrate for qualitative infrared analysis.
Alfa Aesar™ Disposable IR Cards, 5x10cm, NaCl window, 19mm aperture
Provide a conveniently mounted, non-absorbing substrate for qualitative infrared analysis.
Thermo Scientific™ iD5 ZnSe ATR for Nicolet™ iS5 FT-IR Spectrometer
High-performance singlebounce ZnSe ATR sample accessory for FTIR. Ideal for fast, reproducible sampling of solids, liquids, films, pastes, or gels.
Alfa Aesar™ Disposable IR Cards
Cardboard card that fits into the holder of a spectrophotometer
Alfa Aesar™ Disposable IR Cards, 5x10cm, KBr window, 19mm aperture
Provide a conveniently mounted, non-absorbing substrate for qualitative infrared analysis.
Collector™ II Diffuse Reflectance Accessory
Thermo Scientific™ Collector II Diffuse Reflectance Accessory is designed for high sensitivity analysis powder samples and gems.
Bel-Art™ SP Scienceware™ Wilmad Rotor and Caps for Bruker MAS Probe Available on GSA/VA Contract for Federal Government customers only.
Eliminate the need to solvate with MAS rotors manufactured from the highest purity Zirconia. Bel-Art SP Scienceware™ Wilmad MAS Rotors provide the ultimate solution for analysis of solid samples. Available for most present solid state NMR spectrometers.
Bel-Art™ SP Scienceware™ Wilmad™ Aldrich FT-NMR Library with Spectral Viewer on CD-ROM Available on GSA/VA Contract for Federal Government customers only.
Allows for Spectra processing using Zoom, Peak Integration and Peak Labeling. The Bel-Art SP Scienceware™ Wilmad™ Aldrich FT-NMR Library with Spectral Viewer on CD-ROM is an electronic book that contains high-resolution 300MHz 1H and 75MHz 13C spectra, integral, chemical structure and technical data for the more than 11,800 compounds.
Scientific Cell Company Liquid Filter Set for Testing Photometric Accuracy in UV Range
For testing photometric accuracy (0.75 Abs) in the UV range, according to European Pharmacopoeia.
Scientific Cell Company Liquid Filter Set for Testing Wavelength Accuracy
For testing wavelength accuracy, according to European Pharmacopoeia.
  spinner