Spectrophotometer Filters
Scientific Cell Company Hellma Analytics Potassium Chloride Filter
Liquid filter for testing stray light, according to Ph. Eur.
Scientific Cell Company Hellma Analytics Ultrapure Water Filter
Reference filter for testing to stray light according to Ph. Eur.
Scientific Cell Company Hellma Analytics Perchloric Acid
Reference filter for testing the photometric accuracy.
  spinner