Spectrophotometers
Thermo Scientific™ NanoDrop™ 2000/2000c Spectrophotometers Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ NanoDrop 2000 and 2000c are full-spectrum, UV-Vis spectrophotometers used to quantify and assess purity of DNA, RNA, Protein and more.
Thermo Scientific™ NanoDrop™ 8000 Spectrophotometer Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ NanoDrop 8000 is a muti-sample micro-volume UV-Vis spectrophotometer.
Thermo Scientific™ NanoDrop™ Lite Spectrophotometer
The Thermo Scientific™ NanoDrop Lite Spectrophotometer is a compact, personal UV-Vis microvolume spectrophotometer that complements the full-featured NanoDrop 2000/2000c and NanoDrop 8000 instruments.
Lovibond™ EC 2000 PT-CO EComparator
Provides an easy way to transition from subjective visual measurement to a nonsubjective, accurate electronic measurement.
BioTek™ Epoch™ 2 Microplate Spectrophotometer Available on GSA/VA Contract for Federal Government customers only.
Fully contained workstation offers a variety of data output options including Wi-Fi, Bluetooth and USB flash drive. BioTek Epoch 2 Microplate Spectrophotometer is a compact, monochromator-based microplate spectrophotometer combines modern touchscreen technology with excellent performance for UV-Vis measurements in 6- to 384-well microplates, cuvettes and in micro-volume samples with the available Take3™ plate.
Thermo Scientific™ Evolution™ 201/220 UV-Visible Spectrophotometers
Empower your analysis and bring versatility and productivity to your lab with the new Thermo Scientific™ Evolution 201 and 220 UV-Vis systems.
Thermo Scientific™ Evolution™ Spectrophotometer: PROMO
2011 R&D 100 Award Winner. Empower your analysis and bring versatility and productivity to your lab with the new Thermo Scientific™ Evolution 201 and 220 UV-Vis systems.
Thermo Scientific™ SPECTRONIC™ 200 Spectrophotometer
Thermo Scientific™ SPECTRONIC™ 200 is the new standard for rugged, simple-to-use visible spectrophotometers for teaching and routine applications.
Thermo Scientific™ SPECTRONIC™ 200 Spectrophotometer
Thermo Scientific™ SPECTRONIC™ 200 is the new standard for rugged, simple-to-use visible spectrophotometers for teaching and routine applications.
Lovibond™ LC 100 Hand-Held Imaging Spectrophotometer
Creates conformance reports in either PDF or Excel formats that can easily be shared with PC or compatible printer
Lovibond™ Colorimeter Tablet Reagent: DPD No. 1
Use with MC500, MD600, SC450, MD100 colorimeters, SP600 spectrophotometer, Pool Control 9, AquaPRO3 Pool and AquaPRO6 Pool photometers and Scuba IIElectronic Pool Tester
Thermo Scientific™ Life Science Analyzer: BioMate™ 3S Spectrophotometer
Your busy life science laboratory demands performance and flexibility — count on the Thermo Scientific™ BioMate 3S UV-Visible spectrophotometer to deliver.
Thermo Scientific™ Evolution™ 260 Bio UV-Visible Spectrophotometer
The Thermo Scientific™ Evolution 260 spectrophotometer offers reliable, high-quality performance and enhanced customer usability.
Thermo Scientific™ Evolution™ 60S UV-Visible Spectrophotometer
The Thermo Scientific™ Evolution 60S UV-Visible spectrophotometer offers exceptional performance and flexibility for routine to research applications.
Thermo Scientific™ Life Science Analyzer: BioMate™ 3S Spectrophotometer, Six-position cell holder
Your busy life science laboratory demands performance and flexibility — count on the Thermo Scientific™ BioMate 3S UV-Visible spectrophotometer to deliver.
Thermo Scientific™ Orion™ AquaMate 8000 UV-Vis Spectrophotometer
Equip your lab with the accurate, easy to use Thermo Scientific™ Orion™ AquaMate 8000 UV-Vis Spectrophotometer to ensure the ideal instrument for water analysis and beyond
Thermo Scientific™ Evolution™ Array UV-Vis Spectrophotometer Accessories
For use with Evolution Array UV-Vis Spectrophotometer
Thermo Scientific™ Evolution™ 260 Bio UV-Visible Computer Control Spectrophotometer: PROMO
The Thermo Scientific™ Evolution 260 spectrophotometer offers reliable, high-quality performance and enhanced customer usability.
Thermo Scientific™ Evolution™ 220 UV-Vis Pharma Value Package
Ensure accurate quality standards with less worry and keep product moving out the door using UV-Vis Pharma Value Packages.
Thermo Scientific™ Evolution™ 201 UV-Vis Pharma Value Package
Ensure accurate quality standards with less worry and keep product moving out the door using UV-Vis Pharma Value Packages.
Thermo Scientific™ Orion™ AquaMate 7000 Vis Spectrophotometer
Choose the accuracy, dependability and ease of use of the Thermo Scientific™ Orion™ AquaMate 7000 Vis Spectrophotometer to optimize your water and wastewater testing
BioTek™ Epoch™ R Microplate Spectrophotometer Available on GSA/VA Contract for Federal Government customers only.
BioStack and 3rd party automation capability available.
BioTek™ Epoch™ Microplate Spectrophotometer Available on GSA/VA Contract for Federal Government customers only.
Offers budget-friendly, advanced detection capabilities. BioTek Epoch Microplate Spectrophotometer is an optical reader which provides precise UV/Vis measurements in one compact, highly versatile unit.
BioTek™ Integration Kit for Epoch™ Microplate Spectrophotometer
Accessory for use with BioTek Epoch Microplate Spectrophotometer (cat. no. 11120570).
Lovibond™ PFXi-880 Series Spectrophotometers PROMO
With RCMSi technology (Remote Calibration & Maintenance Service) for remote calibration and servicing via internet without leaving benchtop
Lovibond™ RT300 Series Portable Spectrophotometers
Provide absolute and difference measurements in various colorimetric systems
Lovibond™ EC 3000 ASTM EComparator
Provides an easy way to transition from subjective visual measurement to a non-subjective, accurate electronic measurement.
Lovibond™ RT500 Series Portable Spectrophotometers
Designed to give fast, precise and accurate color measurement information on various colorimetric systems
Lovibond™ Colorimeter Solution Reagent: SPADNS
For use with Fluoride Aqua Comparator Test Kit, SC450, MD100 Fluoride colorimeters, MC500, MD600 Multi-Parameter Colorimeters and SP600 Spectrophotometer.
Lovibond™ EC 3000 Saybolt EComparator
Provides an easy way to transition from subjective visual measurement to a non-subjective, accurate electronic measurement.
  spinner