Transilluminators
UVP High-Performance UV Transilluminators
Exclusive 25-watt tubes provide high intensity performance and uniform UV illumination plus sensitivity on stained gels
Spectroline™ Select™ Series UV Transilluminators
Economically priced transilluminators with high-clarity fluorescent response, sensitivity to 1 to 2ng, 312nm wavelength, variable filter areas and Outstanding irradiance uniformity.
UVP White Light Transilluminator
Supplying back-lit white light
UVP Benchtop UV Transilluminators: 3UV
New design features upgraded components for increased efficiency, performance and functionality
UVP UV/White Light 8-Watt Transilluminators
Newly-designed for better performance and efficiency
UVP Benchtop UV Transilluminators: Single UV
New design features upgraded components for increased efficiency, performance and functionality
UVP Model M-10E Mini UV Transilluminators
Small footprint conserves valuable bench space
UVP UV Light Converter Plates
Plate is specially designed to easily fit on the transilluminator filter surface
UVP Filter Assemblies for White Light Transilluminators
Replacement filters for TW-26 and TW-43 models white light transilluminators
Dynamic Diagnostics Wee Sight™ Transilluminator
Helps to accurately locate the tiny veins in a neonate's small limbs
UVP Model M-10E Mini UV Transilluminators: Filter Assembly
Accessory for UVP Model M-10E Mini UV Transilluminators
Spectroline™ Bi-O-Vision™ Series White Light Transilluminators
Offers both medium wave UV (312nm) and white light for dual-light convenience in one box
Spectroline™ Slimline™ UV Transilluminators
Compact models are ideal for photodocumentation of small electrophoresis gels
UVP Benchtop UV Transilluminators: Single UV, Variable Intensity
New design features upgraded components for increased efficiency, performance and functionality
Hoefer™ Replacement UV Lamp
300nm, 6W (UV-B) lamp for use in the UVTM-10 transilluminator
UVP Visi-Blue™ Transilluminators
UV-safe units convert 365nm UV to 460-470nm blue light for use with various stains including GFP stains as well as SYBR Green, and SYPRO Orange stains. The amber protective cover blocks blue light transmission and allows visualization of most stains above 500nm.
UVP Benchtop UV Transilluminators: 2UV
New design features upgraded components for increased efficiency, performance and functionality
Edvotek™ Midrange UV Transilluminator Products for Science Education
Midrange (302nm) UV transilluminator designed to visualize DNA stained with ethidium bromide.
UVP UV/White Light 8-Watt Transilluminator Products for Science Education
UVP UV/White Light 8-Watt Transilluminator is newly-designed for better performance and efficiency.
  spinner