Turbidity Meter Standards and Calibrators
Thermo Scientific™ Remel™ McFarland Equivalence Turbidity Standards Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel McFarland Equivalence Turbidity Standard is packaged in plastic case with a visual comparison card.
Thermo Scientific™ Remel™ Colorimeter Standard Set, Vitek™ Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel Colorimeter Standard Kit contains latex equivalence turbidity standards for use with Vitek colorimeter.
GFS Chemicals Amco Clear™ Linear Calibration Sets
Highly accurate, stable and reproducible standards for calibrating turbidimeters and nephelometers.
GFS Chemicals Amco Clear™ Individual Primary Turbidity Standards
Highly accurate, stable and reproducible standards for calibrating turbidimeters and nephelometers.
Lovibond™ Calibration Set for Infrared Lab Turbidimeter
For use with TB300-IR Lab Turbidimeter.
GFS Chemicals Amco Clear Primary and Deluxe Turbidity Standards Kits
Highly accurate, stable and reproducible standards for calibrating turbidimeters and nephelometers. GFS Chemicals Amco Clear Primary and Deluxe Turbidity Standards Kits are the only alternative to formazin formally approved as a primary standard by the U.S. EPA.
HF Scientific™ ProCal™ Turbidity Standard Reagents Kit for Hach™ 2100AN Turbidimeter
Standards not harmful to the environment or to worker safety
Supply Solutions StablCal™ Certified Low-Level Verification Standards
Precise, pre-mixed Formazin solutions needs no preparation or dilutions
LaMotte™ Turbidity Standards Available on GSA/VA Contract for Federal Government customers only.
For use with LaMotte Portable Turbidity Meters
LaMotte™ Turbidity Standards Products for Science Education
For use with LaMotte Portable Turbidity Meters
  spinner