Consistometers
CSC Bostwick Consistometer
Accurately determines sample consistency
Cannon™ Fenske Routine Viscometers, CFRC Series
For measuring kinematic viscosity of transparent Newtonian liquids, particularly petroleum products or lubricants, according to ASTM D 445 and ISO 3104.
Cannon™ Zeitfuchs™ Cross-Arm Viscometers, ZCAC-RO/ZCAXC-RO Series
Measures kinematic viscosity of transparent and opaque Newtonian liquids according to ASTM D 445 and ISO 3104.
  spinner