Glass Capillary Viscometers
Fisherbrand™ Glass Kinematic Viscometer Tubes
Cannon™-Fenske type viscometer tubes for measuring kinematic viscosity of petroleum products by ASTM™ D 445.
Fisherbrand™ Glass Ubbelohde Calibrated Viscometer Tubes
For measurement of kinematic viscosities of petroleum products according to ASTM™ D 445 and D 446.
Fisherbrand™ Glass Opaque Calibrated Viscometer Tubes
Cannon™-Fenske opaque type viscometer tubes for measuring viscosity of dark liquids and lubricating oils by ASTM™ D 445.
Cannon™ Ubbelohde Calibrated UBC Series Viscometers
Suspended level instrument for measurement of kinematic viscosity.
Cannon™ Zeitfuchs Cross-Arm Viscometer
Use to determine kinematic viscosities of transparent and opaque Newtonian liquids according to ASTM D 445 and ISO 3104.
Fisherbrand™ Glass Vacuum Viscometer Tubes
Cannon™-Manning type viscometer tubes for measuring viscosities of highly viscous materials such as asphalt cement at 60°C (140°F) according to ASTM™ D 2171.
Fungilab™ Cannon-Fenske Opaque Capillary Viscometers
Fungilab Cannon-Fenske Opaque Capillary Viscometers are used for dark Newtonian liquids and to study shearing stress and shearing rate.
Fungilab™ ISO 2431 Flow Cup Viscometers
Measure the viscosity of paint, ink, varnishes and similar products industries. ISO 2431 Flow Cup Viscometer comes in a variety of sizes.
Cannon™ Ubbelohde™ Calibrated CUSMC Series Semi-Micro Viscometers
For measuring kinematic viscosity of small samples of transparent Newtonian liquids according to ASTM D 445 and ISO 3104.
Cannon-Manning Calibrated CMSMC Semi-Micro Viscometers
For measurement of kinematic viscosity of small samples of transparent Newtonian liquids, including petroleum products or lubricants, according to ASTM D 445 and ISO 3104.
Cannon™ Fenske Opaque Viscometer
CFO Calibrated Series that determines viscosities using ASTM testing methods.
Cannon™ Ubbelohde Calibrated CUC Series Viscometers
Suspended level possessing same viscometer constant at all temperatures.
Cannon™ Fenske Calibrated CFOC Series Opaque Viscometers
Measures kinematic viscosity of transparent and opaque Newtonian liquids according to ASTM D445, ISO 3104 and ASTM D2170.
Fungilab™ ASTM D-1200 Flow Cup Viscometers
Measure the viscosity of paint, ink, varnishes, and similar products industries. ASTM D-1200 Flow Cup Viscometers are available in a vareity of sizes.
Koehler™ Instrument Round Holder for Viscometers Available on GSA/VA Contract for Federal Government customers only.
For use with Ubbelohde glass capillary viscometers.
  spinner