Syringe Filters
Thermo Scientific™ Target2™ PVDF Syringe Filters Available on GSA/VA Contract for Federal Government customers only.
Ensure cleaner sample extracts by removing interfering materials and fine particles prior to injection with these robust PVDF syringe filters.
EMD Millipore™ Millex™-GP Sterile Syringe Filters with PES Membrane Available on GSA/VA Contract for Federal Government customers only.
Designed for sterilizing and clarifying sterile solutions. EMD Millipore Millex™-GP Sterile Syringe Filters with PES Membrane are ready-to-use, syringe-driven units with a unique membrane that results in six times less protein binding. Available in a medical grade unit, with a PES or acrylic tip.
EMD Millipore™ Millex™ Sterile Syringe Filters: Durapore™ PVFD Membrane Available on GSA/VA Contract for Federal Government customers only.
Designed for sterilizing and clarifying sterile solutions. EMD Millipore Millex™ Sterile Syringe Filters: MCE Membrane are ready-to-use, syringe-driven units that have never been touched by human hands during filter assembly. Feature 0.1, 0.22, 0.45 and 5.0µm pore sizes; medical-grade filters available.
EMD Millipore™ Millex™ Nonsterile 33mm Syringe Filters Available on GSA/VA Contract for Federal Government customers only.
Remove particulates, reduce signal-to-noise ratios and maintain flat baselines. EMD Millipore Millex™ Nonsterile 33mm Syringe Filters are constructed of polyethylene housing with Durapore™ membranes for preparing samples for analysis. Available in two pore sizes.
EMD Millipore™ Millex™ Nonsterile Syringe Filters with PE Housing - Nylon Available on GSA/VA Contract for Federal Government customers only.
For removing particulates, reducing signal-to-noise ratios and maintaining flat baselines
Fisherbrand™ Syringe Filters: PTFE Membrane Available on GSA/VA Contract for Federal Government customers only.
Low hold-up volume helps you to recover virtually all of your sample after filtering
Corning™ SFCA Syringe Filters Available on GSA/VA Contract for Federal Government customers only.
High burst strength for assured performance
GE Healthcare Whatman™ GD/X Glass Micro Fiber (GMF) Syringe Filters Available on GSA/VA Contract for Federal Government customers only.
Processes three to seven times more sample volume than filters without prefilter. Whatman™ GD/X Glass Micro Fiber (GMF) Syringe Filters are specifically designed for filtration of viscous or otherwise hard-to-filter samples with high solids content.
Fisherbrand™ Syringe Filters - Sterile, Nylon;Sterile; Pore size: 0.2μm; Dia.: 25mm Available on GSA/VA Contract for Federal Government customers only.
Low hold-up volume helps you to recover virtually all of your sample after filtering
Corning™ PES Syringe Filters Available on GSA/VA Contract for Federal Government customers only.
Manufactured from polyethersulfone (PES) for the fastest flow rates, low protein binding and low extractables. Corning™ PES Syringe Filters are ideal for tissue culture and HPLC applications.
Thermo Scientific™ Target2™ Nylon Syringe Filters Available on GSA/VA Contract for Federal Government customers only.
Ensure cleaner sample extracts by removing interfering materials and fine particles prior to injection with these robust nylon syringe filters. Ideal for general HPLC use.
EMD Millipore™ Millex™ Sterile Syringe Filters: MCE Membrane Available on GSA/VA Contract for Federal Government customers only.
Designed for sterilizing and clarifying sterile solutions. EMD Millipore Millex™ Sterile Syringe Filters: MCE Membrane are ready-to-use, syringe-driven units that have never been touched by human hands during filter assembly. Feature 0.22, 0.45 and 0.80µm pore sizes; medical-grade filters available.
GE Healthcare Whatman™ GD/X PVDF Syringe Filters Available on GSA/VA Contract for Federal Government customers only.
Processes three to seven times more sample volume than filters without prefilter. Whatman™ GD/X PVDF Syringe Filters are specifically designed for filtration of viscous or otherwise hard-to-filter samples with high solids content.
GE Healthcare Whatman™ Puradisc 25mm Syringe Filters: Sterile Available on GSA/VA Contract for Federal Government customers only.
Purifies aqueous and organic solutions. Whatman™ Puradisc 25mm Syringe Filters have polypropylene housings with female luer lock inlet and male luer outlet.
GE Healthcare Whatman™ GD/X PTFE Syringe Filters Available on GSA/VA Contract for Federal Government customers only.
Processes three to seven times more sample volume than filters without prefilter. Whatman™ GD/X PTFE Syringe Filters are specifically designed for filtration of viscous or otherwise hard-to-filter samples with high solids content.
Fisherbrand™ Syringe Filters: Nylon Membrane Available on GSA/VA Contract for Federal Government customers only.
Low hold-up volume helps you to recover virtually all of your sample after filtering
Fisherbrand™ Syringe Filters - Nonsterile, Durapore™ PVDF Membrane Available on GSA/VA Contract for Federal Government customers only.
Low hold-up volume helps you to recover virtually all of your sample after filtering
GE Healthcare Whatman™ Mini-UniPrep™ Syringeless Filters Available on GSA/VA Contract for Federal Government customers only.
Processes sample loads in one-third the time. Whatman™ Mini-UniPrep™ Syringeless Filters provide a faster and easier way to remove particulates from samples being prepared for HPLC and UHPLC analysis.
EMD Millipore™ Automation Compatible Filters Available on GSA/VA Contract for Federal Government customers only.
Trouble-free performance on a variety of automated systems
GE Healthcare Whatman™ GD/X Nylon Syringe Filters Available on GSA/VA Contract for Federal Government customers only.
Processes three to seven times more sample volume than filters without prefilter. Whatman™ GD/X Nylon Syringe Filters are specifically designed for filtration of viscous or otherwise hard-to-filter samples with high solids content.
GE Healthcare Whatman™ GD/X Sterile PES Syringe Filters Available on GSA/VA Contract for Federal Government customers only.
Processes three to seven times more sample volume than filters without prefilter. Whatman™ GD/X Sterile PES Syringe Filters are specifically designed for filtration of viscous or otherwise hard-to-filter samples with high solids content.
Corning™ Nylon Syringe Filters Available on GSA/VA Contract for Federal Government customers only.
Recommended for applications requiring very low extractables because they do not contain any wetting agents, detergents or surfactants. Corning™ Nylon Syringe Filters provide excellent chemical resistance for optimum performance in filtering aggressive solutions.
GE Healthcare Whatman™ Puradisc Nylon Syringe Filters: Nonsterile Available on GSA/VA Contract for Federal Government customers only.
Caters to a wide variety of sample types and applications, including capillary electrophoresis, biological preparations, protein filtration, and UV spectroscopy sample preparation. Whatman™ Puradisc Nylon Syringe Filters: Nonsterile are resistant to a range of organic solvents and suitable for use with high pH samples.
EMD Millipore™ Millex™ Sterile Syringe Filters: PES Membrane - Clear Available on GSA/VA Contract for Federal Government customers only.
Ready-to-use syringe-driven units, designed for sterilizing and clarifying sterile solutions
GE Healthcare Whatman™ GD/X Glass Fiber (GF) Syringe Filters Available on GSA/VA Contract for Federal Government customers only.
Processes three to seven times more sample volume than filters without prefilter. Whatman™ GD/X Glass Fiber (GF) Syringe Filters are specifically designed for filtration of viscous or otherwise hard-to-filter samples with high solids content.
GE Healthcare Whatman™ Anotop™ 25 Sterile Syringe Filters Available on GSA/VA Contract for Federal Government customers only.
Remove virtually 100% of particulates larger than pore size
Sartorius™ Minisart™ NY Syringe Filters: Nonsterile Available on GSA/VA Contract for Federal Government customers only.
For rapid small volume filtration of both aqueous and solvent-based samples with maximum user convenience.
GE Healthcare Whatman™ Puradisc™ 25mm Polypropylene Syringe Filters Available on GSA/VA Contract for Federal Government customers only.
Whatman™ Puradisc™ 25mm Polypropylene Syringe Filters may be used for biological sample and HPLC aqueous sample preparation.
Sartorius™ Minisart™ Plus Syringe Filters Available on GSA/VA Contract for Federal Government customers only.
Includes a glass fiber pre-filter for difficult to filter liquids. Sartorius™ Minisart™ Plus Syringe Filters are single-use, complete filtration units with a surfactant-free cellulose acetate membrane. Choose from three color-coded pore sizes in sterile or non-sterile options.
  spinner