Gas Tight Syringes
Hamilton™ 1000 Series Gastight™ Syringes
Ideal for despensing both liquids and gases
Trajan™ eVol and eVol NMR Digital Analytical Syringe Accessories
Charger for eVol™ digitally controlled positive displacement dispensing system
Hamilton™ 1000 Series Gastight™ Syringes: Luer Lock Syringes, TLL Termination Edge
Mid-volume GASTIGHT syringe ideal for dispensing volumes from 100 μL up to 100 mL. 
Restek™ Presents Hamilton Standard Microliter Volume Syringes: Needle: N
Available with cemented needles or removable needles.
Hamilton™ 1000 Series Gastight™ Syringes: Fixed-Needle Syringes, LTN Termination, 22 Gauge, 2 in. Needle Length Edge
Mid-volume GASTIGHT syringe ideal for dispensing volumes from 100 μL up to 100 mL. A variety of different terminations and needle options are available in this series.
Hamilton™ Neuros™ 7000 Series Modified Microliter Syringes with Knurled Hub Needle
An industry first, the Neuros accurately dispenses 50nL to 100μL of liquid through an ultrafine needle protected by a custom-designed sleeve. The Neuros minimizes injection site damage by increasing needle rigidity and delivering a microvolume of liquid to an exact location.
Hamilton™ 1700 Series Gastight™ Syringes: RN Termination Edge
Gastight version of the original 700 series syringes, ideal for dispensing volumes from 1 up to 500μL
Hamilton™ 1000 Series Gastight™ Syringes: Removable-Needle, RN Termination, 22 Gauge, 2 in. Needle Length Edge
Mid-volume GASTIGHT syringe ideal for dispensing volumes from 100 μL up to 100 mL.
Hamilton™ 1700 Series Gastight™ Syringes: N Termination Edge
Gastight version of the original 700 series syringes, ideal for dispensing volumes from 1 up to 500μL
Hamilton™ Modified 7000 Microliter™ Syringes
This series of syringes is ideal for dispensing volumes from 0.05 µL up to 5 µL. The 7000 series is the only zero dead volume syringe. The fine gauge plunger wire travels to the tip of the needle ensuring that all solvent is expelled from the needle.
Hamilton™ Neuros™ 1700 Series Gas Tight Syringes with Removable Needle
An industry first, the Neuros accurately dispenses 50nL to 100μL of liquid through an ultrafine needle protected by a custom-designed sleeve. The Neuros minimizes injection site damage by increasing needle rigidity and delivering a microvolume of liquid to an exact location.
Thermo Scientific™ Fixed Needle Autosampler Syringes for Agilent Instruments Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Fixed Needle Autosampler Syringes for Agilent Instruments are accurate and reproducible over a range of volumes.
Hamilton™ 800 Microliter™ Syringes, RN Termination
Attached to the flange of the 700 series syringe is a blue Syringe Holder that provides a spot to hold the syringe that will not heat up the barrel causing dispense inaccuracies. Additionally it provides support to small volume plungers that are prone to bending during injection. This series of syringes is ideal for dispensing volumes from 0.5 µL up to 250 µL.
Hamilton™ 1700 Series Gastight™ Syringes: LTN Termination Edge
Gastight version of the original 700 series syringes, ideal for dispensing volumes from 1 up to 500μL
Hamilton™ 1700 Series Gastight™ Syringes: LT Termination
Gastight version of the original 700 series syringes, ideal for dispensing volumes from 1 up to 500μL
Hamilton™ 700 Microliter™ Syringes: RN Termination
The 700 series syringe is the original hand-fitted Hamilton syringe. This series of syringes is ideal for dispensing volumes from 0.5 µL up to 500 µL. A variety of different terminations and needle options are available in this series.
Hamilton™ 1800 Series Gastight™ Fixed-Needle Syringes
Reinforced plunger version of the 1700 series syringes is ideal for dispensing volumes from 1 μL up to 500 μL.
Hamilton™ 1700 and 1000 Series Gastight™ SampleLock™ Syringes, 22 Gauge
Hamilton SampleLock syringes allow you to collect, store, transport and analyze liquid or gaseous samples without the danger of evaporative loss or environmental contamination.
Hamilton™ 1700 Series Gastight™ Syringes: TLL Termination
Gastight version of the original 700 series syringes, ideal for dispensing volumes from 1 up to 500μL
Hamilton™ 700 Microliter™ Syringes: LT Termination
The 700 series syringe is the original hand-fitted Hamilton syringe. This series of syringes is ideal for dispensing volumes from 0.5 µL up to 500 µL. A variety of different terminations and needle options are available in this series.
Hamilton™ 700/1700 Series Microliter/Gastight™ Syringes
Hamilton Company offers the most comprehensive selection of standard and custom syringes in the industry. Each is manufactured to achieve the highest level of accuracy and precision possible.
SGE™ Gastight Syringes: Removable Luer-Lok Models
Suitable for liquid and gas samples
Hamilton™ 700 Microliter Syringes: N Termination
The 700 series syringe is the original hand-fitted Hamilton syringe. This series of syringes is ideal for dispensing volumes from 0.5 µL up to 500 µL. A variety of different terminations and needle options are available in this series.
Hamilton™ 1800 Series Gastight™ Removable-Needle Syringes
Reinforced plunger version of the 1700 series syringes is ideal for dispensing volumes from 1 μL up to 500 μL.
Hamilton™ 1000 Series Gastight™ Purge and Trap GC Syringes
Designed for the analysis of drinking water samples according to EPA purge and trap concentration techniques
Hamilton™ 700/700 Microliter™: KH Termination
Syringes have a built-in syringe guide, designed to prevent the plunger from bending and pulling out of the syringe
  spinner