Portable Chlorine Meters
LaMotte™ Chlorine TRACER PockeTester
TRACER PockeTester Total Chlorine Kit, Read Total Chlorine from 0.00 to 9.99ppm to 0.01ppm resolution, Extra bold display includes an analog bar graph feature, uses four LR-44 batteries, Readings are not affected by sample color
LaMotte™ Chlorine TRACER PockeTester Products for Science Education
TRACER PockeTester Total Chlorine Kit, Read Total Chlorine from 0.00 to 9.99ppm to 0.01ppm resolution, Extra bold display includes an analog bar graph feature, uses four LR-44 batteries, Readings are not affected by sample color
  spinner