Indicating Half Cell Electrodes
Thermo Scientific™ Orion™ 900100 Sure-Flow™ Reference Half-Cell Electrode
Precisely measure with ion-selective electrodes using this sleeve-type Ag/AgCl reference electrode.
Thermo Scientific™ Orion™ 900200 Sure-Flow™ Reference Half-Cell Electrode
Perform ISE determinations with the Thermo Scientific™ Orion™ 900200 Sure-Flow™ Reference Half-Cell Electrode, a double junction reference electrode.
Thermo Scientific™ Orion™ 8101BNWP ROSS™ Half-Cell Electrode
Get results three to five times more precise than those obtained with conventional electrodes with the Thermo Scientific™ Orion™ 8101BNWP ROSS™ Half-Cell Electrode.
Fisherbrand™ Glass Body Ag/AgCl Reference Electrodes - Mercury-Free
Recommended for stability at higher temperatures and for use with strongly basic samples
Fisherbrand™ accumet™ Glass Body Mercury-Free Reference Electrodes
Mercury-free, refillable, TRIS-compatible reference electrodes
Mettler Toledo™ InLab™ Reference Half Cells Available on GSA/VA Contract for Federal Government customers only.
For routine applications and measurements in ion-selective and pH half-cells. Mettler Toledo InLab InLab Reference Half Cells Protects against organic and corrosive samples and is ideal for optimum silver ion-free accuracy at various sample temperatures.
Thermo Scientific™ Orion™ Low Resistance pH Electrode, with BNC waterproof connector
Use the Thermo Scientific™ Orion™ 9162BNWP Low Resistance pH Electrode in viscous solutions.
Mettler Toledo™ InLab™ Mono pH Half Cell Electrodes Available on GSA/VA Contract for Federal Government customers only.
For applications where life time of the pH electrode is significantly shorter than the reference electrode.
Fisherbrand™ accumet™ ISE and pH Reference Electrodes - Mercury - Free
Fisherbrand™ accumet™ ISE and pH Reference Electrodes - Mercury - Free is specially designed for superb stability with ion-selective electrodes. It contains a tough, unbreakable, chemical-resistant olymer body and has a high flowrate that is an excellent choice for many pH and titration applications.
Fisherbrand™ accumet™ Metallic ORP Indicating Electrode - Mercury - Free
Platinum Indicating Half-Cell for ORP measurements and redox titrations
Mettler Toledo™ Metal Half Cell Electrodes Available on GSA/VA Contract for Federal Government customers only.
For electrochemical applications.
Fisherbrand™ accumet™ Epoxy Body Mercury-Free Reference Electrode
A rugged epoxy body makes this a great choice for demanding environments
Thermo Scientific™ Orion™ 9101S pH Half-Cell Electrode
Get accurate, reproducible results with the Thermo Scientific™ Orion™ 9101S pH Half-Cell Electrode.
Thermo Scientific™ Orion™ 800300 ROSS™ Sure-Flow™ Half-Cell
Rapidly and accurately measure dirty or viscous samples that would clog traditional electrodes with the Thermo Scientific™ Orion™ 800300 ROSS™ Sure-Flow™ Half-Cell.
  spinner