Micro pH Electrodes
Thermo Scientific™ Orion™ PerpHecT™ ROSS™ Combination pH Micro Electrode
Measure micro volume samples with this refillable, glass double junction electrode.
Thermo Scientific™ Orion™ 9810BN Micro pH Electrode
Measure pH as low as 0.5μL samples in gels or micro containers with this small-tipped glass combination electrode for pH measurements in micro volume samples.
Thermo Scientific™ Orion™ 9863BN Micro pH Electrode
Measure pH by piercing septa with the Thermo Scientific™ Orion™ 9863BN Micro pH Electrode, a bevel needle-tipped 16-gauge glass body combination electrode.
Thermo Scientific™ Orion™ 9826BN Micro pH Electrode
Accurately measure pH in NMR cells with the Thermo Scientific™ Orion™ 9826BN Micro pH Electrode.
OHAUS™ Starter Series Electrodes: Micro pH Electrodes Products for Science Education
Use in combination with Starter bench and portable meters. OHAUS Starter Series Electrodes: Micro pH Electrodes can be used for testing samples in a tube, where diameter of the electrode should be smaller than the tube.
  spinner