pH Electrode Accessories
Thermo Scientific™ Orion™ pH Electrode Cleaning and Storage Solutions
Restore proper electrode performance and prolong electrode life using Thermo Scientific™ Orion™ pH Electrode Cleaning and Storage Solutions.
Thermo Scientific™ Orion™ pH Electrode Filling Solution
Use Thermo Scientific™ Orion™ pH Electrode Filling Solution with pH electrodes and reference electrodes.
Filling Solution: Saturated KCl, 29.8% Available on GSA/VA Contract for Federal Government customers only.
For calomel single-junction electrodes; outer chamber of Ag/AgCl double-junction electrodes; Fluoride, Sodium Combination ISEs, accupHast and accuTupH electrode outer chambers
Filling Solution: 4M KCl saturated with AgCl
For single-junction Ag/AgCl electrodes, Calcium Combination ISEs, inner chamber of many double-junction electrodes
Mettler Toledo™ KCl 3M Electrode Filling Solution Available on GSA/VA Contract for Federal Government customers only.
Equipped with a special cap to facilitate easy filling of electrodes. Mettler Toledo™ KCl 3M Electrode Filling Solution is an electrolyte solution that enables optimum performance of pH and ion-selective electrodes. This bridge electrolyte contains no silver ions than can contaminate samples.
Mettler Toledo™ InLab™ Sensor Cables: MultiPin™ Connection
Electrode cables to connect Mettler Toledo InLab sensors to any Mettler Toledo meter.
Mettler Toledo™ Buffer Solutions: Standard pH Available on GSA/VA Contract for Federal Government customers only.
Storage and refill solutions for pH electrodes
Thermo Scientific™ Accessories for Orion™ pH Electrodes
Store and protect your pH electrodes.
Mettler Toledo™ InLab™ Sensor Cables: S7 Connection
Electrode cables to connect Mettler Toledo InLab sensors to any Mettler Toledo meter.
Thermo Scientific™ Orion™ Electrode Adapter Cables
Convert an electrode with one connector to the required connector of another meter by using the Thermo Scientific™ Orion™ Electrode Adapter Cables.
Fisherbrand™ Filling Solution for accuCap™ Electrode
For Capillary Junction Electrode 13-620-130
Ricca Chemical Electrode Storage Solution
For storing glass and combination pH electrodes.
Thermo Scientific™ Orion™ Storage Sleeve and Base
Protect electrodes from breakage when not in use with the Thermo Scientific™ Orion™ Electrode Storage Sleeve and Base.
Mettler Toledo™ Electrode Cable for Coulometric Karl Fischer™ Titrator
Accessory for use with Coulometric Karl Fischer Titrators.
Fisherbrand™ accumet™ Electrode Reservoir - Mercury - Free
Reservoir, Electrode; Fisher Scientific accumet; Stores one electrode in appropriate soaking solution; Oval, polyethylene bottle; Leakproof seal; 1L x 2W x 3.75 in. H
Oakton™ pH Electrode Cleaning Solution
Remove grease and oil deposits from your electrodes with this cleaning solution. Oakton pH Electrode Cleaning Solution contains pepsin.
Fisherbrand™ Electrode Connection Adapters - Mercury - Free
Adapters connect electrodes to various input jacks and are mercury free
Mettler Toledo™ Thiourea Cleaning Solution
Removes silver sulfide contamination from pH electrodes. Mettler Toledo™ Thiourea Cleaning Solution helps maintain electrode performance and should be utilized after measurements and prior to recalibration. Treatment should be applied until discoloration.
Mettler Toledo™ Electrode Fill Solutions Available on GSA/VA Contract for Federal Government customers only.
Made from the highest quality reagents to provide optimum performance.
ACROS Organics™ Electrode Storage Solution
Store pH electrodes to make them last
Mettler Toledo™ InLab™ Sensor Cables: For Reference Electrodes
Electrode cables to connect Mettler Toledo reference electrodes to any meter.
Fisherbrand™ accumet™ 12mm Karl Fisher Electrode Bulb Protector
Protect fragile glass membrane against breakage
Thermo Scientific™ Orion™ Electrode Extension Cables
Extend the length of an electrode cable by 15′ (4.57m) when using the Thermo Scientific™ Orion™ Electrode Extension Cables.
Mettler Toledo™ pH Electrode Accessories: Glass Flow Cell
For use with electrode with 12mm dia. shaft
  spinner