Digital Thermometers
Fisher Scientific™ Gun/Fold-Up Thermometers Products for Science Education
Provides wide measurement range with Rotary probe design.
Fisher Scientific™ Water-resistant Thermometer Products for Science Education
Use this thermometer to measure temperature of water.
Fisher Scientific™ Traceable™ Hi-Accuracy Refrigerator Thermometer with Min/Max Readings Products for Science Education
Stores in memory and displays minimum/maximum readings for last 24 hours (each hour), seven days (each day) and 30 days (each day). Fisher Scientific Traceable Hi-Accuracy Refrigerator Thermometers include visual LEDs and audible alarms which signal continuously even if temperature returns to non-alarm range.
Fisher Scientific™ Traceable™ Memory-Card Refrigerator/Freezer Thermometers/Humidity Meters Products for Science Education
Min/max memories, high/low alarms and data capture; displays °F/°C. Fisher Scientific Traceable Memory-Card Refrigerator/Freezer Thermometers/Humidity Meters capture a continuous stream of data to see the behavior of refrigerators, room areas or tests.
Fisher Scientific™ Pen-Shaped Thermometers Products for Science Education
Measure temperature either in Celcius or Farenheit scale or both.
Fisherbrand™ Traceable™ Hi-Accuracy Thermometers Products for Science Education
Monitors inlet and outlet simultaneously. Fisherbrand Traceable Hi-Accuracy Thermometer records time and date for MIN/MAX readings.
Fisher Scientific™ Traceable™ Lollipop™ Shock/Waterproof Thermometer Products for Science Education
Offers minimum/maximum temperature readings. Fisher Scientific Traceable Lollipop Shock/Waterproof Thermometer is water-resistant and is ideal for wet labs, wash-down areas, and all field applications.
Fisher Scientific™ Hygro-Thermometers Products for Science Education
With external sensor for temperature.
Fisher Scientific™ Pen-Shaped Thermometers: With Data Hold Function Products for Science Education
Measure temperature either in Celcius or Farenheit scale or both.
Fisher Scientific™ Room Thermometer Products for Science Education
Thermometer with data hold feature.
Fisher Scientific™ Traceable™ Big-Digit See-Thru™ Thermometers Products for Science Education
Attach to the outside of a window to view outdoor temperatures. Fisher Scientific Traceable Big-Digit See-Thru Thermometers have transparent display with 1.25 in.H (3.1cm) digits which show current and minimum/maximum temperatures.
Fisher Scientific™ Traceable™ Digital Dial Thermometers Products for Science Education
Easy to read digital dial and mercury free. Fisher Scientific Traceable Digital Dial Thermometers are ideal for cuvets, soil, incubators and test tubes. Wide temperature span combines the ranges of more than 90 different mercury thermometers.
Fisher Scientific™ Traceable™ Digital-Bottle™ Refrigerator/Freezer Thermometers Products for Science Education
Accurately monitor temperature in freezers, refrigerators, incubators and environmental chambers with this temperature-buffered sensor.
Fisher Scientific™ Sentry Thermometers Products for Science Education
Shows current, high and low temperatures simultaneously. Fisher Scientific™ Traceable™ Sentry™ Thermometer instantly shows if water bath, freezer or incubator is within temperature specifications and monitors temperature of any experiment, even when you are not there.
Fisher Scientific™ Traceable™ Memory-Card Humidity/Temperature/Dew Point Products for Science Education
Min/max memories and high/low alarms, plus integrated datalogger. Fisher Scientific Traceable Memory-Card Humidity/Temperature/Dew Point meter captures 5.9 million readings with removable 256MB SD card. Monitors cabinets, incubators, cleanrooms, storerooms, labs, hoods and critical areas.
Fisher Scientific™ Round T-Shaped Thermometers Products for Science Education
Measure temperature either on Celsius or Farenheit scale or both.
Fisher Science Education™ Traceable™ Extreme-Accuracy Standards Thermometer Products for Science Education
Provides the exact readings required for critical requirements, quality control checks and routine measurements. Fisher Science Education Traceable Extreme-Accuracy Standards Thermometers are ideal for measuring temperatures in water baths, cuvets, gas systems, reagents, standards solutions, ovens, incubators, pharmaceuticals, petroleum products, foods and wastewater.
Fisher Scientific™ Jumbo Refrigerator/Freezer Thermometers Products for Science Education
Digital thermometers are perfect for monitoring freezers, water baths, heating blocks, incubators and refrigerators. Fisher Scientific Jumbo Refrigerator/Freezer Thermometer accurately monitors temperature with enclosed “temperature-buffered” sensor.
Fisher Scientific™ Small Handheld Digital Thermometer Products for Science Education
Thermometers with wide measurement range.
Fisher Scientific™ Traceable™ Extra-Extra-Long-Probe Waterproof Thermometer Products for Science Education
20 in. probe for hard-to-reach deep vessels. Fisher Scientific Traceable Extra-Extra-Long-Probe Waterproof Thermometer is ideal for use in large beakers, flasks, vats, pails and drums.
Fisher Scientific™ Traceable™ Digital Thermometer Products for Science Education
Versatile panel-mount module snaps into place or can be used as a stand-alone unit.
Fisher Scientific™ Fridge/Freezer/Room Thermometer Products for Science Education
With freezer/room temperature display.
Fisher Scientific™ Traceable™ Platinum Freezer Thermometer Products for Science Education
Accurately monitors temperatures in freezers, water baths, heating blocks, incubators and refrigerators.
Fisher Science Education™ Traceable™ Hygrometer/Thermometer/Barometer/Dew Point Pen Products for Science Education
User-selectable dual display shows humidity/temperature, humidity/dew point, barometer/temperature or barometer/humidity.
Fisher Scientific™ Traceable™ Wireless Radio-Signal Refrigerator Thermometer Products for Science Education
Wirelessly monitors refrigerators from up to 100 ft. away, offering min/max memory and high/low alarm. Fisher Scientific Traceable Wireless Radio-Signal Refrigerator Thermometer includes a remote refrigerator bottle probe module with three channels (set to 1, 2 and 3).
Fisher Scientific™ T-Shaped Digital Thermometer Products for Science Education
Stainless-steel probe with protective probe cover.
Fisher Scientific™ Tri-Display Thermometer Products for Science Education
Triple-display for current, minimum and maximum temperature.
Fisher Scientific™ Round T-Shaped Thermometers: Data Hold Function Products for Science Education
Measure temperature either on Celcius or Farenheit scale or both.
Fisher Scientific™ Long-Stem Digital Thermometer Products for Science Education
Durable and safe, containing no mercury or alcohol. Fisher Scientific Long-Stem Digital Thermometers are rugged and versatile digital thermometers to read soil, or stir and measure the temperature of solutions. Can also be used with incubators.
  spinner