Benchtop Titrators
Mettler Toledo™ EasyPlus™ Easy Cl Titrators
Used to measure and speeds up simple, automated precipitation titrations. Mettler Toledo EasyPlus Easy Cl Titrators are ideal for determination of chloride content in food industry, salt content of waste and drinking water, sulfide or mercaptan content of oils and lubricants and silver content of solutions and dissolved metals.
Mettler Toledo™ Replacement Parts for C20 Karl Fischer™ Autotitrator Available on GSA/VA Contract for Federal Government customers only.
Accessories For use with Mettler Toledo C20 Karl Fischer™ Autotitrator
Mettler Toledo™ EasyPlus™ Easy Pro Titrators
Combines acid/base inflection point, pH endpoint and precipitation/redox indicative titrations to determine most common parameters in food, beverage and chemical products. Mettler Toledo EasyPlus Easy Pro Titrators are ideal for determination of simple titrations of juice, wine, p- and m-value, free fatty acids in food and alkalinity, chloride in food or water and sulfides in oils.
Mettler Toledo™ V20S Compact Volumetric KF Titrator
Ideal for fast and precise water content determination with easy operation. Mettler Toledo V20S Compact Volumetric KF Titrators can analyze samples with water content between 100 ppm and 100% in single measurements or sample series with statistics.
Mettler Toledo™ EasyPlus™ Easy Ox Titrators
Ideal for determination of routine volumetric Karl Fischer titration in food, chemical products and other sample types. Mettler Toledo EasyPlus Easy Ox Titrators have small design which is capable of performing any basic titration with ease.
Mettler Toledo™ V10S Compact Volumetric KF Titrator
Ideal for fast and precise water content determination with easy operation. Mettler Toledo V10S Compact Volumetric KF Titrators can analyze samples with water content between 100 ppm and 100% in single measurements or sample series with statistics.
Mettler Toledo™ EasyPlus™ Easy pH Titrators
Simplifies and speeds up automated acid/base endpoint or inflection/equivalence point titrations. Mettler Toledo EasyPlus Easy pH Titrators have small design which is capable of performing any basic titration with ease.
Mettler Toledo™ EasyPlus™ Easy KFV Titrators
Ideal for determination of routine volumetric Karl Fischer titration in food, chemical products and other sample types.
Mettler Toledo™ C20S Compact Coulometric KF Titrators Available on GSA/VA Contract for Federal Government customers only.
Ideal for fast and precise water content determination with very simple operation. Mettler Toledo C20S Coulometric KF Titrators can analyze samples with water content between 1ppm and 5% in single measurements or as sample series with statistics.
Mettler Toledo™ G20S Compact Potentiometric Titrator Available on GSA/VA Contract for Federal Government customers only.
Ideal for fast and precise potentiometric titration with very simple operation. Mettler Toledo G20S Compact Potentiometric Titrator has an easy-to-use interface and space saving design which makes it an ideal choice for daily routine analysis.
Mettler Toledo™ C10S Compact Coulometric KF Titrators Available on GSA/VA Contract for Federal Government customers only.
Ideal for fast and precise water content determination with very simple operation. Mettler Toledo C10S Coulometric KF Titrators can analyze samples with water content between 1ppm and 5% in single measurements or as sample series with statistics.
Mettler Toledo™ G10S Compact Potentiometric Titrator Available on GSA/VA Contract for Federal Government customers only.
Ideal for fast and precise potentiometric titration with very simple operation. Mettler Toledo G10S Compact Potentiometric Titrator has an easy-to-use interface and space saving design which makes it an ideal choice for daily routine analysis.
LaMotte™ Direct Reading Titrator Available on GSA/VA Contract for Federal Government customers only.
Replacement for glass direct reading titrators
EMD Millipore™ AQUASTAR™ CombiSolvent Keto Solvent
One component volumetric Karl Fischer titration volumetric solvent (Methanol-free) of 1L.
LaMotte™ Direct Reading Titrator Products for Science Education
For applications prohibiting use of glass.
LaMotte™ Direct Reading Titrator Products for Science Education
Replacement for glass direct reading titrators
  spinner