Microtube Racks
Thermo Scientific™ FlipStrip™ Microtube Racks with Lid Available on GSA/VA Contract for Federal Government customers only.
Hold 0.2mL strips and 1.5mL tubes on one side of the Thermo Scientific™ FlipStrip Microtube Rack with Lid, and 0.5mL tubes on the other. Made from 100% recycled plastic.
Thermo Scientific™ 4-Way Flipper™ Racks
Hold thirty-two 0.5mL, thirty-two 1.5mL, twelve 15mL or four 50mL tubes, each on a different side, using Thermo Scientific™4-Way Flipper™ Racks.
Thermo Scientific™ Reversible Microtube Racks with Lid Available on GSA/VA Contract for Federal Government customers only.
Store samples securely and rapidly identify them with Thermo Scientific™ Reversible Microtube Racks with Lid: featuring a low-profile design with an alphanumeric grid.
Thermo Scientific™ Nalgene™ Polypropylene Floating Microtube Racks Available on GSA/VA Contract for Federal Government customers only.
Thaw, cool and incubate samples at elevated or reduced temperatures. These floating microtube racks are especially useful in genetic cloning, microbiology and immunology labs.
Thermo Scientific™ Nalgene™ Microcentrifuge Tube Storage Boxes Available on GSA/VA Contract for Federal Government customers only.
Safely store samples using Thermo Scientific™ Nalgene™ Microcentrifuge Tube Storage Boxes.
Thermo Scientific™ Nalgene™ Microcentrifuge Tube Rack: ResMer™ Manufacturing Technology Available on GSA/VA Contract for Federal Government customers only.
Save lab space with these chemical-resistant tube racks. Stackable racks have molded letters and numbers for easy identification.
Fisherbrand™ Beta Blocks
Designed to hold 1.5mL microcentrifuge tubes
Thermo Scientific™ 96-Well Flipper™ Microtube Racks
Protect samples and allow racks to be stacked using this 96-Well Flipper Microtube Rack.
Thermo Scientific™ Owl™ RF-3 Filing System
Organize your refrigerator, coldroom, freezer or benchtop with this filing system.
Bel-Art™ SP Scienceware™ Round Bubble Racks Available on GSA/VA Contract for Federal Government customers only.
Optimized for use in microbiology and immunology labs
Bel-Art™ SP Scienceware™ Styrene Microcentrifuge Tube Ice Racks
For temperature-stabilized benchtop procedures or storage
Fisherbrand™ Microcentrifuge Tube Racks
Two frosted writing areas for sample identification
Bel-Art™ SP Scienceware™ PrepSafe™ Microcentrifuge Tube Mini Floating Racks Available on GSA/VA Contract for Federal Government customers only.
Immobilizing tabs protect tubes from opening when under pressure, submerged or knocked over
Fisherbrand™ Microcentrifuge Tube Rack
For 1.5mL micro test tubes
Bel-Art™ SP Scienceware™ Prepsafe Microcentrifuge Tube Mini Floating Rack Accessory
Vortexing attachment for samples to be vortexed simultaneously.
Fisherbrand™ Polypropylene Microtube Support Racks Available on GSA/VA Contract for Federal Government customers only.
Excellent for microbiology work or any work using microtubes
Axygen™ PCR Tube Storage Racks Available on GSA/VA Contract for Federal Government customers only.
Constructed of polypropylene. Corning™ Axygen&trade PCR Tube Storage Racks hold 96 0.2mL PCR tubes. Supplied with lids.
Fisherbrand™ Polypropylene Microtube Storage Racks
Instantly organize microcentrifuge test tubes in any lab
Bel-Art™ SP Scienceware™ Microcentrifuge Tube Ice Racks/Tray Available on GSA/VA Contract for Federal Government customers only.
Deep tray holds ice and water for extended use time
Bel-Art™ SP Scienceware™ No-Wire™ Microcentrifuge Tube Racks
Open sides optimize use in crushed ice or water baths
Bel-Art™ SP Scienceware™ 5mL Centrifuge Tube Rack
For 5mL centrifuge tubes, including Eppendorf brand, this rack will also accommodate other tubes with diameter of 13-16mm
Fisherbrand™ 96-Well PCR Racks
Designed for preparing and storing 0.2mL thin-walled PCR tubes and strip tubes before or after PCR sample processing applications
Connecting Microcentrifuge Tube Racks Products for Science Education
Maximize storage and save benchtop space with microcentrifuge tube racks that are stackable when loaded or empty
Bel-Art™ SP Scienceware™ Poxygrid™ 50mL Centrifuge Tube Racks Products for Science Education Available on GSA/VA Contract for Federal Government customers only.
Organizes tubes for storage and processing. Bel Art™ SP Scienceware™ Poxygrid™ 50mL Centrifuge Tube Racks.
Fisherbrand™ Acrylic Rack Holder
Maximizes benchtop or freezer space
  spinner