Cell Culture Tubes
PYREX™ Disposable Round-Bottom Rimless Glass Tubes Available on GSA/VA Contract for Federal Government customers only.
Manufactured from borosilicate glass to reduce pH changes and contaminants. Corning™ PYREX™ Disposable Round-Bottom Rimless Glass Tubes are designed for both tissue culture and general bacteriological work.
PYREX™ VISTA™ Rimless Culture Tubes Products for Science Education
Offer convenient plugging and rack storage.
Kimble™ Disposable Glass Culture Tube
Round bottomed with threaded neck
Kimble™ ValueWare™ Culture Tubes With Marking Spots Products for Science Education
With fire-polished top rim and uniform wall for maximum heat transfer and chemical resistance.
PYREX™ VISTA™ Rimless Reusable Glass Culture Tubes Available on GSA/VA Contract for Federal Government customers only.
Functionality, economy, and quality
Kimble™ KIMAX™ Culture Tube Screw Cap and Rubber Liner Products for Science Education
Convenient spot on tube for marking identification.
Kimble™ Rim Top Mark-M™ Borosilicate Glass Culture Tubes with Marking Labels Available on GSA/VA Contract for Federal Government customers only.
Permanent ceramic label, fused to the glass located 3/4 in. from the open end
Fisherbrand™ Reusable Glass Rim Top Culture Tubes Available on GSA/VA Contract for Federal Government customers only.
Uniform wall thickness for maximum heat transfer and chemical resistance
Fisherbrand™ Disposable Auto-Analyzer Cups Available on GSA/VA Contract for Federal Government customers only.
Specifically designed for use with most clinical instrumentation
PYREX™ Culture Tubes with Threaded End and Marking Spot Available on GSA/VA Contract for Federal Government customers only.
For tissue culture and general bacteriological work
Fisherbrand™ Disposable Borosilicate Glass Tubes with Plain End Available on GSA/VA Contract for Federal Government customers only.
Use these tubes for tissue culture and clinical chemistry applications. Fisherbrand™ Disposable Borosilicate Glass Tubes with Plain End are made of borosilicate glass to reduce pH changes and contaminants potentially leached from soda lime glass.
PYREX™ Disposable Rimless Flat-Bottom Glass Tubes Available on GSA/VA Contract for Federal Government customers only.
Ideal for plant tissue culture and general culture work
Pyrex™ Round Bottom Threaded Culture Tubes Available on GSA/VA Contract for Federal Government customers only.
Made from borosilicate glass for use with samples sensitive to leaching from plastic tubes. Corning™ Pyrex™ Round Bottom Threaded Culture Tubes are designed for tissue culture and general bacteriological work.
Kimble™ ValueWare™ Culture Tubes with Rubber Lined Phenolic Caps Products for Science Education
Mineral-filled cap to withstand repeated autoclaving.
PYREX™ VISTA™ Reusable Glass Tubes with Phenolic Screw Caps Available on GSA/VA Contract for Federal Government customers only.
Supplied with deep-form phenolic caps to facilitate handling and sealing after autoclaving
Kimble™ Kontes™ Screw Thread Culture Tubes with Unattached Rubber Lined Caps
Uniform wall thickness for maximum heat transfer and heat resistance
Fisherbrand™ False Bottom Vials
Facilitate the automation process with an orientation identifier and an indicator for bar code label placement
Starplex Scientific™ False Bottom Analyzer Tube Available on GSA/VA Contract for Federal Government customers only.
Designed for high-volume analyzer aspiration probes incapable of reaching the bottoms of larger sample tubes
Simport™ Scientific Sample Tubes for Roche Cobas Analyzers
Polystyrene tubes offer excellent clarity for easy viewing of contents
Globe Scientific Analyzer Specific Sample Cup
Made to exact specifications, these sample cups provide excellent optical properties
Evergreen Scientific BMC/Hitachi™ 717, 736, 737 Analyzer Sample Cups
For use with BMC/Hitachi Analyzer models 717, 736, and 737
Oak Ridge False Bottom Analyzer Tubes
Tubes are sealed with a screw-on cap
  spinner