Color Comparison Tubes
DWK Life Sciences Kimble™ Color Comparison APHA Standard Tubes Available on GSA/VA Contract for Federal Government customers only.
High-quality glass test tubes that meet Federal and American Public Health Association (APHA) standards
DWK Life Sciences Kimble™ Low-Form Color-Comparison Tubes Available on GSA/VA Contract for Federal Government customers only.
Shadowless bottom construction allows for viewing with undistorted light
DWK Life Sciences Kimble™ Low Form Nessler Color Comparison Tubes Available on GSA/VA Contract for Federal Government customers only.
Fill Kimble™ Low Form Nessler Color Comparison Tubes and let their shadowless bottom transmit undistorted light. No dark spots form when the tubes are filled with liquid and viewed from the top.
Lovibond™ Model 711-A Nesslerizer System Nessler Tubes
Set of two clear glass Nessler tubes
  spinner