Environmental Sampling Vials
Thermo Scientific™ Clear VOA Glass Vials with 0.125in. Septa
Filling vials without air entrapment is effortless due to the perfect shoulder angle of these VOA vials which are offered in a wide range of sizes and styles.
Thermo Scientific™ TOC Certified Containers
Trust our low-level certified vials for Total Organic Carbon testing and sampling. TOC-certified products are manufactured, packaged and tested to exacting standards.
Thermo Scientific™ Amber VOA Glass Vials with Closed-Top Cap
Designed for EPA volatile organic analysis (VOA) when a pierceable septa closure is not needed.
Thermo Scientific™ National Assembled Convenience Kits: EPA Vials with Caps and Septa Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ National EPA Vial Convenience Kits are designed for discrete water sampling applications.
Thermo Scientific™ Amber VOA Glass Vials with 0.125in. Septa
Filling vials without air entrapment is effortless due to the perfect shoulder angle of these VOA vials which are offered in a wide range of sizes and styles.
Thermo Scientific™ Clear VOA Glass Vials with Closed-Top Cap
Thermo Scientific™ Clear VOA Vials with Closed Cap are designed for EPA volatile organic analysis (VOA).
Thermo Scientific™ Premium Pack Amber Glass VOA Vials with 0.125in. Septa
Protect your most critical samples from collection to analysis. Featuring special foam cell dividers and assembled dust covers to protect the injection surface prior to use.
Thermo Scientific™ Amber VOA Glass Vials with 0.060in. Septa
Optimized for automation, these vials feature a thinner septum that is easier to puncture, and is permanently bonded to the cap to prevent unseating during analysis.
Thermo Scientific™ Premium Pack Clear Glass Vials with 0.125in. Septa
Protect your most critical samples from collection to analysis. Featuring special foam cell dividers and assembled dust covers to protect the injection surface prior to use.
Thermo Scientific™ Depyrogenated Sterile Empty Vials
Thermo Scientific™ Sterile Empty Vials are offered in capacities ranging from 1 to 100mL.
Thermo Scientific™ Silanized Glass Vials
Save time and minimize waste costs when performing quantitative analysis or storing materials by using these pretreated, silanized vials.
Thermo Scientific™ National Components for Vial Convenience Kits Available on GSA/VA Contract for Federal Government customers only.
Vials, caps and septa for use with Thermo Scientific™ National Vial Convenience Kits.
Thermo Scientific™ Economy Certified Glass VOA Vials with 0.125in. Septa
Designed for water sample collection for volatile organic analysis, these vials are available in the standard 40mL size and feature all the essentials and none of the frills.
Thermo Scientific™ Premium Pack Clear Glass Vials with 0.060in. Septa
Protect your most critical samples from collection to analysis. These vials feature protective packaging, and a thinner septum that is optimized for automation.
Thermo Scientific™ Capitol Vial EPA Vials
Patented flip top with permanent hinge lid
Thermo Scientific™ Economy Processed VOA Glass Vials with Septa
Keep costs low while eliminating the need to wash vials in-house. Vials are processed the same way as our certified VOA vial, but come without certification documentation.
Thermo Scientific™ Amber Clean Snap Vials with 0.125in. Septa
Protect samples from contamination during storage or transport with Clean Snap Vials. An innovative cap protects the septum and provides convenient access at time of analysis.
Thermo Scientific™ Premium Pack Amber Glass Vials with Closed-Top Cap, 20mL
Protect critical samples from collection to analysis. Special foam cell dividers holds vials securely separated, each in their own individual cell for ultimate vial protection.
Thermo Scientific™ Clear Clean Snap Vials with 0.125in. Septa
Protect samples from contamination during storage or transport with Clean Snap Vials. An innovative cap protects the septum and provides convenient access at time of analysis.
Thermo Scientific™ Clear VOA Glass Vials with 0.060in. Septa
Optimized for automation, these vials feature a thinner septum that is easier to puncture, and is permanently bonded to the cap to prevent unseating during analysis.
Fisherbrand™ Economical Glass EPA Vials
Ideal for use in water sampling
Fisherbrand™ EnviroWare™ Assembled Borosilicate Glass EPA Vials
For the collection and storage of water samples
Fisherbrand™ Glass EPA Vials
Packaged in partitioned trays
Kimble™ Kontes™ Screw Thread Vials
Combine a low coefficient of expansion with a very high chemical resistance
Kimble™ EPA Water Analysis Screw Thread KG-33 Borosilicate Glass
Clear and amber screw thread EPA vials for the collection and storage of water samples
Kimble™ EPA Water Analysis Vials
Ensure proper collection and storage of water samples
Wheaton™ Glass EPA Vials Available on GSA/VA Contract for Federal Government customers only.
Choose clear or amber borosilicate glass vials
  spinner