Vacuum Pumps
Thermo Scientific™ Direct-Drive Vacuum Pumps Available on GSA/VA Contract for Federal Government customers only.
Ideally suited for vacuum drying, filtration, impregnation, extraction, concentration, centrifugation, freeze-drying, degassing and cryogenic studies
Fisher Scientific™ Maxima™ C Plus Vacuum Pump
Pumps provide convenient deep vacuum for demanding lab equipment applications
EMD Millipore Millivac™ Mini Pump Available on GSA/VA Contract for Federal Government customers only.
Compact, maintenance-free pumps that provide a consistent source of vacuum for filtration and other laboratory applications
Fisher Scientific™ MaximaDry™ Oil-Free Vacuum Pumps, Two Stage Available on GSA/VA Contract for Federal Government customers only.
Clean, reliable, oil-free vacuum pumps are ideal for use with rotary evaporators, gel dryers, filtration flasks and manifolds, vacuum ovens and concentrators
Fisher Scientific™ Air Cadet™ Vacuum/Pressure Station
Features built-in bronze, brass vacuum/pressure gauge and stainless-steel regulator
Fisher Scientific™ Air Cadet™ Single-Head Vacuum/Pressure Pumps
Pumps are economically priced and easily handle tough vacuum/pressure applications
EMD Millipore High Output Pumps Available on GSA/VA Contract for Federal Government customers only.
Features a piston-driven design for greater power
Fisher Scientific™ MaximaDry™ Oil-Free Vacuum Pumps, One Stage Available on GSA/VA Contract for Federal Government customers only.
Clean, reliable, oil-free vacuum pumps are ideal for use with rotary evaporators, gel dryers, filtration flasks and manifolds, vacuum ovens and concentrators
Fisher Scientific™ MaximaDry™ Oil-Free Vacuum Pumps, Parallel Stage
Clean, reliable, oil-free vacuum pumps are ideal for use with rotary evaporators, gel dryers, filtration flasks and manifolds, vacuum ovens and concentrators
EMD Millipore Chemical Duty Vacuum Pressure Pump Available on GSA/VA Contract for Federal Government customers only.
Decrease process filtration times. EMD Millipore Chemical Duty Vacuum Pressure Pump supports high flow rates for faster pumping. Cast-aluminum pumphead, housing, and regulator with optional chemical-resistant internal components. Light weight and portable so they can be shared among workstations.
Heidolph™ Valve-Regulated Vacuum Pumps - Rotavac Available on GSA/VA Contract for Federal Government customers only.
Standard vacuum pumps can be controlled manually or via valve operated vacuum controllers
Labconco™ VACUUBRAND™ Hybrid™ Vaccum Pumps
Integrates high performance rotary vane pump with a chemical-resistant diaphragm pump to create a low maintenance, longer-lasting pump
Labconco™ Rotary Vane Vacuum Pumps
Designed for use with Labconco Freeze-dry Systems—compact size easily fits into freeze-dry system cabinets
Labconco™ Scroll Vacuum Pump Accessory, Silencer
For use with Labconco™ Scroll vacuum pump. Labconco™ Scroll Vacuum Pump Accessory, Silencer combines filtration efficiency with noise reduction.
Heidolph™ RPM-Regulated Vacuum Pumps - Rotavac Available on GSA/VA Contract for Federal Government customers only.
For use with all Heidolph Hei-VAP models
Labconco™ Diaphragm Vacuum Pumps
For use with CentriVap™ Centrifugal Concentrators, RapidVap™ Vacuum Evaporation Systems and Vacuum Desiccators
Labconco™ Scroll Vacuum Pump
For use with FreeZone™ Freeze Dry Systems - Oil-free, dry pumps requiring very little maintenance. Labconco™ Scroll Vacuum Pump is dry pump that use two spiral scrolls to compress air and vapors and move them toward exhaust.
Heidolph™ Hei-VAC Vacuum Pump Available on GSA/VA Contract for Federal Government customers only.
For all Heidolph Hei-VAP models
Lift Pump Products for Science Education
Demonstrate how a well pump works.
Vacuum Pumps Products for Science Education
Compact and lightweight for easy portability.
BrandTech™ VACUUBRAND™ NT Diaphragm Vacuum Pumps: ME4 NT Available on GSA/VA Contract for Federal Government customers only.
For a wide variety of non-corrosive and non-evaporative applications. ME-series pumps feature an ultimate vacuum of 52 Torr.
BrandTech™ VACUUBRAND™ MZ2C NT Synchro Dry Chemistry Multitasking Vacuum System Available on GSA/VA Contract for Federal Government customers only.
Provides powerful oil-free vacuum for two independent applications, such as rotary evaporators, gel dryers or other common lab tasks.  VACUUBRAND™ MZ2C NT Synchro Dry Chemistry Multitasking Vacuum System builds on the popular MZ2C NT diaphragm vacuum pump.
International Vacuum Pumps Products for Science Education
Remove moisture and air from tanks and other containers with hand held vacuum pump.
BrandTech™ VACUUBRAND™ NT Diaphragm Vacuum Pumps: ME8 NT Available on GSA/VA Contract for Federal Government customers only.
For a wide variety of non-corrosive and non-evaporative applications. ME-series pumps feature an ultimate vacuum of 52 Torr.
  spinner