Dialysis Tubing
Thermo Scientific™ SnakeSkin™ 3.5K MWCO Dialysis Tubing
Perform desalting and buffer exchange for 10 to 100mL samples with this 3500-MW cut-off (MWCO), ready-to-use form of traditional dialysis membrane tubing.
Thermo Scientific™ SnakeSkin™ 10K MWCO Dialysis Tubing
Perform desalting and buffer exchange for 10 to 100mL samples with this 10,000-MW cut-off (MWCO), ready-to-use form of traditional dialysis membrane tubing.
Seamless Cellulose Dialysis Tubing Products for Science Education
Retains materials with 12,000 and higher molecular weight.
Thermo Scientific™ SnakeSkin™ Dialysis Tubing Clips
Securely close dialysis tubing up to 50mm flat diameter, including SnakeSkin Dialysis Tubing, with these sturdy, non-floating tube closures.
Thermo Scientific™ SnakeSkin™ 7K MWCO Dialysis Tubing
Perform desalting and buffer exchange for 15 to 100mL samples with this 7000-MW cut-off (MWCO), ready-to-use form of traditional dialysis membrane tubing.
Fisherbrand™ Regenerated Cellulose Dialysis Tubing
Diffuses low molecular weight compounds in aqueous solutions; retains proteins and bacteria in tubing
Spectrum™ Spectra/Por™ 7 Membrane Tubing (Sulfur and Heavy Metal Free):1 1000 to 50,000 Dalton MWCO. Available on GSA/VA Contract for Federal Government customers only.
Ideal for molecular separation, protein binding studies, electrolyte removal, desalting
Spectrum™ Labs Spectra/Por™ 5 12-14 kD MWCO Standard RC Dry Dialysis Kits
With Standard Grade Regenerated Cellulose membrane tubing for applications with significant size difference between MW species being separated
Spectrum™ Trial Size Kits for Biotech-Grade CE Dialysis Membrane Tubing (1 meter)
Ideal for small, critical dialysis applications that require the more concise selectivity and retention provided by Biotech Cellulose Ester (CE) Membrane than Standard Grade RC
Spectrum™ Labs Spectra/Por™ 6 50,000 D MWCO Standard RC Pre-wetted Dialysis Kits
With Pre-wetted ready-to-use tubing, not requiring soaking to remove glycerol
Spectrum™ Spectra/Por™ 6 Pre-wetted Standard RC Dialysis Tubing, 1-50 kD MWCO Available on GSA/VA Contract for Federal Government customers only.
Clear, flexible and sturdy; Standard Grade RC (regenerated cellulose) membrane conveniently pre-equilibrated in water, eliminating the need to soak to remove glycerin and hydrate the membrane
Spectrum™ Labs Spectra/Por™ 1 6-8 kD MWCO Standard RC Dry Dialysis Kits
With Standard Grade Regenerated Cellulose membrane tubing for applications with significant size difference between MW species being separated
Spectrum™ Labs 300 kD Biotech CE Dialysis Kits
With tubing made from biologically inert and ultra-pure Biotech cellulose ester membrane
Spectrum™ Labs Spectra/Por™ 6 3500 D MWCO Standard RC Pre-wetted Dialysis Kits
With Pre-wetted ready-to-use tubing, not requiring soaking to remove glycerol
Spectrum™ Labs 3.5 - 5 kD Biotech CE Dialysis Kits
With tubing made from biologically inert and ultra-pure Biotech cellulose ester membrane
Spectrum™ Labs 1000 kD Biotech CE Dialysis Kits
With tubing made from biologically inert and ultra-pure Biotech cellulose ester membrane
Spectrum™ Labs Spectra/Por™ 6 25 kD MWCO Standard RC Pre-wetted Dialysis Kits
With Pre-wetted ready-to-use tubing, not requiring soaking to remove glycerol
Spectrum™ Labs 0.5 - 1.0 kD Biotech CE Dialysis Kits
With tubing made from biologically inert and ultra-pure Biotech cellulose ester membrane
Spectrum™ Spectra/Por™ 4 RC Dialysis Membrane Tubing 12,000 to 14,000 Dalton MWCO Available on GSA/VA Contract for Federal Government customers only.
Ideal for molecular separation, protein binding studies, electrolyte removal, desalting
Spectrum™ Labs Spectra/Por™ 7 10,000 D MWCO Standard RC Pre-Treated Dialysis Kits
With chemically-treated tubing to remove any trace levels of heavy metals and sulfide compounds
Spectrum™ Labs Spectra/Por™ 2 and 4 12-14 kD MWCO Standard RC Dry Dialysis Kits
For smaller dialysis experiments or evaluating dialysis tubing for dialysis application
Spectrum™ Trial Size Kits for Biotech-Grade RC Dialysis Membrane Tubing
0.5 meter (1.6 ft) dialysis tubing length ideal for small applications or membrane evaluation
Spectrum™ Labs Spectra/Por™ 7 25,000 D MWCO Standard RC Pre-Treated Dialysis Kits
With chemically-treated tubing to remove any trace levels of heavy metals and sulfide compounds
Spectrum™ Labs Spectra/Por™ 7 2000 D MWCO Standard RC Pre-Treated Dialysis Kits
With chemically-treated tubing to remove any trace levels of heavy metals and sulfide compounds
Spectrum™ Labs 50 kD Biotech CE Dialysis Kits
With tubing made from biologically inert and ultra-pure Biotech cellulose ester membrane
Spectrum™ Labs 3.5 - 5 kD Biotech RC Dialysis Kits
With tubing made from Biotech RC membrane that can tolerate concentrated-weak acids, bases, dilute-strong acids, bases, most alcohols and some mild or dilute organic solvents
Spectrum™ Spectra/Por Biotech-Grade RC Dialysis Membrane Tubing
A premium dialysis membrane that provides superior molecular selectivity than Standard RC and superior tolerance than Biotech CE
Spectrum™ Labs 8 - 10 kD Biotech CE Dialysis Kits
With tubing made from biologically inert and ultra-pure Biotech cellulose ester membrane
Spectrum™ Spectra/Por™ 2 RC Dialysis Membrane Tubing 12,000 to 14,000 Dalton MWCO Available on GSA/VA Contract for Federal Government customers only.
Ideal for molecular separation, protein binding studies, electrolyte removal, desalting
  spinner