Rubber Stoppers
DWK Life Sciences Kimble™ Kontes™ Hollow White Rubber Sleeve Stoppers Available on GSA/VA Contract for Federal Government customers only.
Hollow stopper fits into the neck of the glassware apparatus. Series 774261 may be used at up to 125°C for up to 8 hours.
Wheaton™ Flange Style Rubber Stopper
For research and parenteral drug applications
Fisherbrand™ Turnover Septum Stoppers
For narrow-aperture containers
Fisherbrand™ Solid Rubber Stoppers
Available in a variety of sizes ranging from 000 through 16
Wheaton™ Straight Plug Style Silicone Stoppers for Vials and Serum Bottles
For maximum sealing of vials with straight wall glass finish
Wheaton™ Ultra Pure Straight Plug Stoppers Available on GSA/VA Contract for Federal Government customers only.
Provides for extremely high chemical purity and low gas permeability. Wheaton™ Ultra Pure Straight Plug Stoppers are manufactured from an ultra pure bromobutyl formulation.
Fisherbrand™ Solid Neoprene Rubber Stoppers
Recommended where rubber would be attacked or deteriorated
DWK Life Sciences Wheaton™ Snap-on Style Stoppers for Vials and Serum Bottles
For maximum sealing of vials with blow-back glass finish
Wheaton™ Lyophilized Stopper: 3-Leg Lyophilization Stopper Available on GSA/VA Contract for Federal Government customers only.
Very low adsorption of the preservatives present in aqueous parenteral solutions - Available in 2-Leg, 3-Leg and Igloo styles.
Wheaton™ Ultra Pure Complete Coat Stoppers Available on GSA/VA Contract for Federal Government customers only.
Featuring PTFE-like coatings to prevent reactions due to sample contact with rubber. Wheaton™ Ultra Pure Complete Coat Stoppers are manufactured in a silicone-free environment.
Fisherbrand™ Twistit™ Rubber Stoppers
Stoppers made from soft, resilient, natural rubber with safe convenient countersunk holes, stoppers feature excellent aging characteristics with high resistance to deformation
Fisherbrand™ Two-Hole Twistit™ Rubber Stoppers
Stoppers made from soft, resilient, natural rubber with safe convenient countersunk holes, stoppers feature excellent aging characteristics with high resistance to deformation
Wheaton™ Straight Plug Style Bromobutyl rubber Stoppers for Vials and Serum Bottles
For maximum sealing of vials with straight wall glass finish
Fisherbrand™ Three-hole Twistit™ Rubber Stoppers
Stoppers made from soft, resilient, natural rubber with safe convenient countersunk holes, stoppers feature excellent aging characteristics with high resistance to deformation
Fisherbrand™ Silicone Recessed Septum Stoppers
Nontoxic and autoclavable recessed septum stoppers are externally serrated to create an effective internal seal
Wheaton™ Lyophilized Stopper: 2-Leg Lyophilization Stopper Available on GSA/VA Contract for Federal Government customers only.
Very low adsorption of the preservatives present in aqueous parenteral solutions - Available in 2-Leg, 3-Leg and Igloo styles.
Kimble™ Kontes™ Pluro Stopper Set Available on GSA/VA Contract for Federal Government customers only.
This autoclavable set of 7 neoprene rubber adapters is designed to fit Buchner and fritted glass funnels.
  spinner