Labware Detergents and Cleaners
Fisherbrand™ Sparkleen™ 1 Detergent
Sparkleen 1 is a universal detergent for manual or hand washing laboratory glassware and equipment. This mild highly buffered product is an excellent soak cleaner for most materials. Glassware washed with Sparkleen 1 is truly sparkling clean, entirely free from washing film, even with the hardest water.
Thermo Scientific™ RBS™ 35 Detergent Concentrate
A no-residue laboratory cleaning solution for labware utensils, surface materials and glassware, including bottles, beakers, pipets, lenses, mirrors.
Decon™ AquaPur™ Purified USP-Grade Water
For use in laboratory or production applications
Fisherbrand™ Sparkleen™ 2 Detergent
Sparkleen 2 effectively cleans the most tenacious soils, such as grease, petrolatum, dried blood and organic deposits quickly and safely. Sparkleen 2 is used for mechanical spray washing of glassware, stainless steel, and most plastics. Providing economical, trouble-free cleaning requiring only minimal attention.
Alconox™ Liquinox™ Critical Cleaning Liquid Detergent
Concentrated, mild, anionic detergent for manual and ultrasonic cleaning
Fisherbrand™ Versa-Clean™ Pharmaceutical Production and Multi-Purpose Cleaner
Fisherbrand Versa-Clean™ From delicate labware to full scale production equipment Versa-Clean keeps sensitive equipment free of inhibitory residues that could interfere with analysis, tissue culture, and other critical procedures. Ideal for pharmaceutical production, research labs, environmental labs, water or wastewater labs.
Alconox™ Powdered Precision Cleaner
Concentrated, anionic detergent for manual and ultrasonic cleaning
Alconox™ Alcotabs™ Critical Cleaning Detergent Tablets
Effervescent detergent tablet for maximum cleaning in syphon pipet washers to clean pipets and tubes
Alconox™ Alcojet™ Low-Foaming Powdered Critical Cleaning Detergent
Concentrated, non-ionic cleaner for use in labware washers, parts washers, power-spray systems, manual and ultrasonic cleaning
Labconco™ LabSolutions™ Powder Detergent
Removes grease, blood, agar, protein digestates, and other contaminants
Alconox™ Tergazyme™ Enzyme-Active Powered Detergent
Concentrated, anionic detergent with protease enzyme for manual and ultrasonic cleaning
Thermo Scientific™ RBS™-pF Detergent Concentrate
A phosphate-free, surfactant-based cleaning solution for use on many laboratory surfaces, labware utensils and glassware.
Decon™ Contrex™ AL Alkaline Liquid Detergent Available on GSA/VA Contract for Federal Government customers only. Get IT Item
For use in manual, soak, or ultrasonic cleaning
Decon™ Contrex™ EZ Powdered Enzymatic Detergent Available on GSA/VA Contract for Federal Government customers only.
Formulated for removal of protein and fat contamination
Labconco™ LabSolutions™ Low-Foaming Liquid Detergent
Removes a wide range of contaminants
Alconox™ Solujet™ Low-Foaming Phosphate-Free Liquid Detergent
Nonionic liquid cleaner for use in labware washers, manual and ultrasonic cleaning
Bel-Art™ SP Scienceware™ Aquet™ Detergent Available on GSA/VA Contract for Federal Government customers only.
For manual cleaning of glassware, plastics and surfaces
Cleaning Solution (Chromic-Sulfuric Acid) Available on GSA/VA Contract for Federal Government customers only.
Removes nearly impossible stains from glassware
Alconox™ Citranox™ Liquid Acid Cleaner and Detergent
Phosphate-free cleaner and metal brightener for high performance removal of metal oxides, scale, salts and inorganic residues in manual and ultrasonic cleaning
MP Biomedicals™ Inc. 7X™ Laboratory Detergents
Safe, effective cleaning solutions for glass and plastic labware
FL-70™ Concentrate Available on GSA/VA Contract for Federal Government customers only.
Powerful alkaline detergent removes dirt, grime, organic residue, coagulated blood, dried gelatin and stopcock grease
Alconox™ Detojet™ Low-Foaming Liquid Detergent
Low-foaming, heavy duty alkaline detergent for use in labware washers, parts washers, power-spray systems and ultrasonic cleaning systems
Alconox™ Citrajet™ Low-Foaming Liquid Acid Cleaner
Liquid cleaner and metal brightener for removal of metal oxides, scales, salts and inorganic residues
Thermo Scientific™ PCC-Pfree™ Detergent Concentrate
A phosphate-free, surfactant-based cleaning solution for use on many laboratory surfaces and glassware; RBS pF alternative for Europe.
  spinner