Surfactants and Wetting Agents
Hexadecyltrimethylammonium bromide, 99+%, ACROS Organics™
CAS: 57-09-0 Molecular Formula: C19H42BrN Molecular Weight (g/mol): 364.456 InChI Key: LZZYPRNAOMGNLH-UHFFFAOYSA-M Synonym: Cetrimonium bromide, Cetyltrimethylammonium bromide, CTABr PubChem CID: 5974 ChEBI: CHEBI:3567 IUPAC Name: hexadecyl(trimethyl)azanium;bromide SMILES: CCCCCCCCCCCCCCCC[N+](C)(C)C.[Br-]
Dodecyl sulfate sodium salt, 99%, for biochemistry, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
CAS: 151-21-3 Molecular Formula: C12H25NaO4S Molecular Weight (g/mol): 288.378 MDL Number: MFCD00036175 InChI Key: DBMJMQXJHONAFJ-UHFFFAOYSA-M Synonym: Dodecyl Sodium Sulfate, SDS, Sodium lauryl sulfate, Sodium lauryl sulfate PubChem CID: 3423265 ChEBI: CHEBI:8984 IUPAC Name: sodium;dodecyl sulfate SMILES: CCCCCCCCCCCCOS(=O)(=O)[O-].[Na+]
Polysorbate 80, Acros Organics™
CAS: 9005-65-6 MDL Number: MFCD00082107 Synonym: Polyoxyethylene(20)sorbitan monooleate
Polysorbate 40, Acros Organics™
CAS: 9005-66-7 Molecular Formula: C26H50O10 Molecular Weight (g/mol): 522.676 MDL Number: MFCD00165345 InChI Key: HMFKFHLTUCJZJO-OQUNMALSSA-N Synonym: Polyoxyethylene (20) Sorbitan Monopalmitate, Polysorbate 40 PubChem CID: 92329579 IUPAC Name: 2-[(2R)-2-[(2R,3R,4R)-3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy]ethyl dodecanoate SMILES: CCCCCCCCCCCC(=O)OCCOCC(C1C(C(CO1)OCCO)OCCO)OCCO
Sodium Lauryl Sulfate (Powder/NF/FCC), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
CAS: 151-21-3 Molecular Formula: C12H25NaO4S Molecular Weight (g/mol): 288.378 MDL Number: MFCD00036175 InChI Key: DBMJMQXJHONAFJ-UHFFFAOYSA-M Synonym: Sodium Dodecyl Sulfate, SDS, Dodecylsulfuric Acid Sodium Salt, Duponol™ C Dry PubChem CID: 3423265 ChEBI: CHEBI:8984 IUPAC Name: sodium;dodecyl sulfate SMILES: CCCCCCCCCCCCOS(=O)(=O)[O-].[Na+]
Tween™ 80, Fisher Chemical
CAS: 9005-65-6 MDL Number: MFCD00082107 Synonym: Polyoxyethylene (20), Polysorbate (80)
Triton™ X-100 (Electrophoresis), Fisher BioReagents™ Available on GSA/VA Contract for Federal Government customers only.
CAS: 9002-93-1 Molecular Formula: C16H26O2 Molecular Weight (g/mol): 250.382 InChI Key: JYCQQPHGFMYQCF-UHFFFAOYSA-N Synonym: Polyethylene Glycol p-tert-Octylphenyl Ether PubChem CID: 5590 IUPAC Name: 2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethanol SMILES: CC(C)(C)CC(C)(C)C1=CC=C(C=C1)OCCO
Sodium Dodecyl Sulfate (SDS), White Powder, Electrophoresis, Fisher BioReagents™
CAS: 151-21-3 Molecular Formula: C12H25NaO4S Molecular Weight (g/mol): 288.378 InChI Key: DBMJMQXJHONAFJ-UHFFFAOYSA-M Synonym: Sodium Lauryl Sulfate, SDS PubChem CID: 3423265 ChEBI: CHEBI:8984 IUPAC Name: sodium;dodecyl sulfate SMILES: CCCCCCCCCCCCOS(=O)(=O)[O-].[Na+]
Triton™ X-100, scintillation grade, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
CAS: 9002-93-1 Molecular Formula: C16H26O2 Molecular Weight (g/mol): 250.382 InChI Key: JYCQQPHGFMYQCF-UHFFFAOYSA-N Synonym: Polyethylene glycol mono [4-(1,1,3,3-tetramethylbutyl)phenyl] ether PubChem CID: 5590 IUPAC Name: 2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethanol SMILES: CC(C)(C)CC(C)(C)C1=CC=C(C=C1)OCCO
Triton™ X-100, 98%, for molecular biology, DNAse, RNAse and Protease free, ACROS Organics™
CAS: 9002-93-1 Molecular Formula: C16H26O2 Molecular Weight (g/mol): 250.382 MDL Number: MFCD00132505 InChI Key: JYCQQPHGFMYQCF-UHFFFAOYSA-N Synonym: Polyethylene glycol mono [4-(1,1,3,3-tetramethylbutyl)phenyl] ether PubChem CID: 5590 IUPAC Name: 2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethanol SMILES: CC(C)(C)CC(C)(C)C1=CC=C(C=C1)OCCO
SDS Solution, 20% Sodium Dodecyl Sulfate Solution, Molecular Biology/Electrophoresis, Fisher BioReagents™ Available on GSA/VA Contract for Federal Government customers only.
CAS: 151-21-3 Molecular Formula: C12H25NaO4S Molecular Weight (g/mol): 288.378 InChI Key: DBMJMQXJHONAFJ-UHFFFAOYSA-M Synonym: Sodium lauryl sulfate, SDS PubChem CID: 3423265 ChEBI: CHEBI:8984 IUPAC Name: sodium;dodecyl sulfate SMILES: CCCCCCCCCCCCOS(=O)(=O)[O-].[Na+]
Alfa Aesar™ Poloxamer 188
CAS: 9003-11-6 Molecular Formula: C5H10O2 Molecular Weight (g/mol): 102.13 MDL Number: MFCD00082049 InChI Key: RVGRUAULSDPKGF-UHFFFAOYNA-N Synonym: Pluronic™ F-68; Polyoxyethylene-polyoxypropylene block copolymer PubChem CID: 24751 IUPAC Name: 2-methyloxirane;oxirane SMILES: CC1CO1.C1CO1
Aerosol™ OT, Anhydrous Solid, Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
CAS: 577-11-7 Molecular Formula: C20H37NaO7S Molecular Weight (g/mol): 444.559 MDL Number: MFCD00012455 InChI Key: APSBXTVYXVQYAB-UHFFFAOYSA-M Synonym: Dioctyl Sulfosuccinate, AOT, DOSS PubChem CID: 23673837 IUPAC Name: sodium;1,4-bis(2-ethylhexoxy)-1,4-dioxobutane-2-sulfonate SMILES: CCCCC(CC)COC(=O)CC(C(=O)OCC(CC)CCCC)S(=O)(=O)[O-].[Na+]
Tween™ 80, Fisher BioReagents
CAS: 9005-65-6 Synonym: Polysorbate 80, Polyoxyethylene-20-sorbitan Monooleate
Tween™ 20, Fisher BioReagents™ Available on GSA/VA Contract for Federal Government customers only.
CAS: 9005-64-5 Molecular Formula: C26H50O10 Molecular Weight (g/mol): 522.676 InChI Key: HMFKFHLTUCJZJO-UHFFFAOYSA-N Synonym: Polysorbate 20, Polyoxyethylene-20-sorbitan Monolaurate, Polyoxyethylenesorbitan monolaurate PubChem CID: 443314 IUPAC Name: 2-[2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy]ethyl dodecanoate SMILES: CCCCCCCCCCCC(=O)OCCOCC(C1C(C(CO1)OCCO)OCCO)OCCO
Sodium Dodecyl Sulfate, Ultrapure Bioreagent, J.T.Baker™
CAS: 151-21-3 Molecular Formula: C12H25NaO4S Molecular Weight (g/mol): 288.378 InChI Key: DBMJMQXJHONAFJ-UHFFFAOYSA-M Synonym: SDS PubChem CID: 3423265 ChEBI: CHEBI:8984 IUPAC Name: sodium;dodecyl sulfate SMILES: CCCCCCCCCCCCOS(=O)(=O)[O-].[Na+]
SDS Solution, 10% Sodium Dodecyl Sulfate Solution, Molecular Biology/Electrophoresis, Fisher BioReagents™
CAS: 151-21-3 Molecular Formula: C12H25NaO4S Molecular Weight (g/mol): 288.378 InChI Key: DBMJMQXJHONAFJ-UHFFFAOYSA-M PubChem CID: 3423265 ChEBI: CHEBI:8984 IUPAC Name: sodium;dodecyl sulfate SMILES: CCCCCCCCCCCCOS(=O)(=O)[O-].[Na+]
n-Decanoylsucrose, ≥97%, Calbiochem™, EMD Millipore™
CAS: 31835-06-0 Molecular Formula: C22H40O12 Molecular Weight (g/mol): 496.55 InChI Key: STVGXWVWPOLILC-UHFFFAOYSA-N Synonym: α-D-Glucopyranoside PubChem CID: 5102725 IUPAC Name: [6-[3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-3,4,5-trihydroxyoxan-2-yl]methyl decanoate SMILES: CCCCCCCCCC(=O)OCC1C(C(C(C(O1)OC2(C(C(C(O2)CO)O)O)CO)O)O)O
Antifoam B™ Silicone Emulsion, BAKER™, J.T.Baker™
A grade of chemicals for laboratory use
Polysorbate 20, Acros Organics™ Available on GSA/VA Contract for Federal Government customers only.
CAS: 9005-64-5 Molecular Formula: C26H50O10 Molecular Weight (g/mol): 522.676 MDL Number: MFCD00165986 InChI Key: HMFKFHLTUCJZJO-UHFFFAOYSA-N Synonym: Polyoxyethylene(20)sorbitan monolaurate PubChem CID: 443314 IUPAC Name: 2-[2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy]ethyl dodecanoate SMILES: CCCCCCCCCCCC(=O)OCCOCC(C1C(C(CO1)OCCO)OCCO)OCCO
Sodium Dodecyl Sulfate (SDS), Micropellets, Fisher BioReagents Available on GSA/VA Contract for Federal Government customers only.
CAS: 151-21-3 Molecular Formula: C12H25NaO4S Molecular Weight (g/mol): 288.378 InChI Key: DBMJMQXJHONAFJ-UHFFFAOYSA-M PubChem CID: 3423265 ChEBI: CHEBI:8984 IUPAC Name: sodium;dodecyl sulfate SMILES: CCCCCCCCCCCCOS(=O)(=O)[O-].[Na+]
ZWITTERGENT™ 3-14 Detergent, Calbiochem™, EMD Millipore™
CAS: 14933-09-6 Molecular Formula: C19H41NO3S Molecular Weight (g/mol): 363.601 InChI Key: BHATUINFZWUDIX-UHFFFAOYSA-N Synonym: n-Tetradecyl-N,N-dimethyl-3-ammonio-1-propanesulfonate PubChem CID: 84705 ChEBI: CHEBI:10129 IUPAC Name: 3-[dimethyl(tetradecyl)azaniumyl]propane-1-sulfonate SMILES: CCCCCCCCCCCCCC[N+](C)(C)CCCS(=O)(=O)[O-]
Triton X-100, MP Biomedicals
CAS: 9002-93-1 Molecular Formula: C16H26O2 Molecular Weight (g/mol): 250.382 InChI Key: JYCQQPHGFMYQCF-UHFFFAOYSA-N PubChem CID: 5590 IUPAC Name: 2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethanol SMILES: CC(C)(C)CC(C)(C)C1=CC=C(C=C1)OCCO
Sodium Lauryl Sulfate, 0.00300 N, Ricca Chemical
CAS: 151-21-3 Molecular Formula: C12H25NaO4S Molecular Weight (g/mol): 288.378 InChI Key: DBMJMQXJHONAFJ-UHFFFAOYSA-M PubChem CID: 3423265 ChEBI: CHEBI:8984 IUPAC Name: sodium;dodecyl sulfate SMILES: CCCCCCCCCCCCOS(=O)(=O)[O-].[Na+]
Dodecyl sulfate sodium salt, 99+%, for molecular biology, DNAse, RNAse and Protease free, ACROS Organics™
CAS: 151-21-3 Molecular Formula: C12H25NaO4S Molecular Weight (g/mol): 288.378 MDL Number: MFCD00036175 InChI Key: DBMJMQXJHONAFJ-UHFFFAOYSA-M Synonym: Dodecyl Sodium Sulfate, SDS, Sodium lauryl sulfate, Sodium lauryl sulfate PubChem CID: 3423265 ChEBI: CHEBI:8984 IUPAC Name: sodium;dodecyl sulfate SMILES: CCCCCCCCCCCCOS(=O)(=O)[O-].[Na+]
Aerosol™ OT Solution, 10% (w/w), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Aerosol™ OT 10% is a remarkable chemical solution that “makes water wetter” because it reduces surface tension and interfacial tension to a considerable degree even at low concentrations
Triton X-100, Alfa Aesar
CAS: 9002-93-1 Molecular Formula: C16H26O2 Molecular Weight (g/mol): 250.382 InChI Key: JYCQQPHGFMYQCF-UHFFFAOYSA-N PubChem CID: 5590 IUPAC Name: 2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethanol SMILES: CC(C)(C)CC(C)(C)C1=CC=C(C=C1)OCCO
Sodium Dodecyl Sulfate (Certified) ACS, Fisher Chemical
CAS: 151-21-3 Molecular Formula: C12H25NaO4S Molecular Weight (g/mol): 288.378 MDL Number: MFCD00036175 InChI Key: DBMJMQXJHONAFJ-UHFFFAOYSA-M Synonym: Sodium Lauryl Sulfate, SDS, Sulfuric Acid Monododecylester Sodium Salt, Irium PubChem CID: 3423265 ChEBI: CHEBI:8984 IUPAC Name: sodium;dodecyl sulfate SMILES: CCCCCCCCCCCCOS(=O)(=O)[O-].[Na+]
Lithium Dodecyl Sulfate (Powder or Flakes/Molecular Biology), Fisher BioReagents Available on GSA/VA Contract for Federal Government customers only.
CAS: 2044-56-6 Molecular Formula: C12H25LiO4S Molecular Weight (g/mol): 272.328 InChI Key: YFVGRULMIQXYNE-UHFFFAOYSA-M PubChem CID: 2735071 IUPAC Name: lithium;dodecyl sulfate SMILES: [Li+].CCCCCCCCCCCCOS(=O)(=O)[O-]
  spinner