Control Environments Sterile Mops
Contec™ VertiKlean™ MAX™ Mopping Systems Mop Heads Available on GSA/VA Contract for Federal Government customers only.
Clean walls, ceilings and other hard-to-reach surfaces with ease. The Contec™ VertiKlean™ MAX™ Mopping Systems Mop Heads feature a variety of heads that easily snap onto frames. The polyurethane foam lasts longer than traditional mops and more evenly releases liquid.
Contec™ Edgeless™ Sterile Mop Heads Available on GSA/VA Contract for Federal Government customers only.
Manufactured from a continuous tube of strong, durable 100% polyester knitted fabric. Contec™ Edgeless™ Sterile Mop Heads are looped to form tubular mop strands that will not shred, break or deteriorate, eliminating contamination.
Contec™ VertiKlean™ Wall Washing System Non-Sterile Mop Heads Available on GSA/VA Contract for Federal Government customers only.
Use with VertiKlean&trade Wall Washing System which is designed for cleaning vertical surfaces. Contec™ VertiKlean™ Wall Washing Systems Non-Sterile Mop Heads and Covers are comprised of laminated polyester fabric and high sorbency polyester foam for excellent cleaning capacity and abrasion resistance. A textured option for applications that require improved particle removal is available.
Contec™ EasyReach™ Cleaning System Available on GSA/VA Contract for Federal Government customers only.
Passes easily through transfer hatches, requires no assembly and is maintenance-free. Contec™ EasyReach™ Cleaning System is ideal for isolators, cabinets, laminar air-flow hoods, glove boxes and other mini environments.
Contec™ VertiKlean™ Wall Washing Systems Sterile Mop Heads Available on GSA/VA Contract for Federal Government customers only.
Use with VertiKlean&trade Wall Washing System which is designed for cleaning vertical surfaces. Contec™ VertiKlean™ Wall Washing Systems Sterile Mop Heads and Covers are comprised of laminated polyester fabric and high sorbency polyester foam for excellent cleaning capacity and abrasion resistance. A textured option for applications that require improved particle removal is available.
Micronova™ T-Mop™ Cleanroom Mops with Covers Available on GSA/VA Contract for Federal Government customers only.
Low-profile, wide-surface mops with covers
Contec™ Edgeless™ Mop Handle with Frame
Integrated handle and frame is convenient and economical. Contec™ Edgeless™ Mop Handle with Frame are designed exclusively for Contec™ Edgeless™ Mop Heads.
Contec™ Polypropylene Double Bucket System Available on GSA/VA Contract for Federal Government customers only.
Support general purpose and controlled environment cleaning with this fully autoclavable system. The Contec™ Polypropylene Double Bucket System features a cart that holds the different colored buckets together securely and a downpress wringer for efficient mop wringing.
Contec™ QuickConect™ Mop Head Frames
Featuring a quick-connect button for easy attachment to all sizes and styles of Contec™ Quick Connect™ handles. Contec™ QuickConnect™ Mop Head Frames are specifically designed for use with the Contec™ family of mopping systems.
Micronova™ SnapMop™: Non-Irradiated Available on GSA/VA Contract for Federal Government customers only.
Can be laundered and autoclaved
Foamtec PharmaMOP™ Cleanroom Mop Heads
Ultra-lightweight large area design for all critical cleanroom surfaces
Contec™ EasyCurve™ Mop Heads
Choose from non-sterile or validated sterile versions; mop heads are autoclavable. The Contec™ EasyCurve™ Mop Heads use curved mop faces with pivoting connectors for easier surface contact at a variety of angles. 
Rubbermaid™ Cleanroom Maintenance Mops Get IT Item
Looped-end, tailbanded mop for greater floor coverage with each stroke
Micronova™ PharmaPly™ Mop Head Available on GSA/VA Contract for Federal Government customers only.
Ideal for covering large surface areas quickly and efficiently
Texwipe™ AlphaMop™ Available on GSA/VA Contract for Federal Government customers only.
Flat mopping system for easy and efficient cleaning of floors, walls and ceilings
Micronova™ NovaPoly™ MicroMop™ Available on GSA/VA Contract for Federal Government customers only.
Die-cut mop efficiently wicks away moisture and is laser cut to minimize particulates
Berkshire™ BCR™ Cleanroom Mopping System: Mop 1
Looped-end construction and fantail design minimize particle generation
Micronova™ SnapMop™: Gamma-Irradiated Available on GSA/VA Contract for Federal Government customers only.
Can be laundered and autoclaved
Contec™ Edgeless™ Non-Sterile Mop Heads Available on GSA/VA Contract for Federal Government customers only.
Manufactured from a continuous tube of strong, durable 100% polyester knitted fabric. Contec™ Edgeless™ Non-Sterile Mop Heads are looped to form tubular mop strands that will not shred, break or deteriorate, eliminating contamination.
Foamtec UltraMOP™ Self-Wringing Mop Heads
Clean room sponge mop head refill for Roll-O-Matic™
Micronova™ J-Zone MegaSorb™ Mop
For applications requiring high absorbency
Micronova™ PolyGen™ Mop Heads Available on GSA/VA Contract for Federal Government customers only.
Economical and low-linting
Micronova™ PowerHead™ Disposable Mop Heads Available on GSA/VA Contract for Federal Government customers only.
Available in several different types and densities of foam and material coverings
Micronova™ MicroMop™ Fabric Mops Available on GSA/VA Contract for Federal Government customers only.
Highly efficient floor mops for cleaning large surface areas quickly
Texwipe™ ClipperMop™ Mop
For use with wet, dry, or sterile wipers
Contec™ MicroGenesis™ Microfiber Wipes Available on GSA/VA Contract for Federal Government customers only.
Pick up and retain particles, dust, and other contaminants in cleanrooms and other critical applications. Contec™ MicroGenesis™ Microfiber Wipes utilize 100% polyester microfiber yarn knitted into a thermally bonded polypropylene substrate. This structure applies microfiber only to the wiping surface where it is needed while sealing the base of each stitch in a thermally bonded base.
  spinner