Controlled Environments Mop Handles
Contec™ QuickConnect™ Mop Handles and Hooks
Choose from a lightweight anodized aluminum, durable electropolished stainless steel or fiberglass. Contec™ QuickConnect™ Mop Handles are available in fixed length or telescoping models to fit all mopping needs.
Contec™ VertiKlean™ Wall Washing System Non-Sterile Mop Heads Available on GSA/VA Contract for Federal Government customers only.
Use with VertiKlean™ Wall Washing System which is designed for cleaning vertical surfaces. Contec™ VertiKlean™ Wall Washing Systems Non-Sterile Mop Heads and Covers are comprised of laminated polyester fabric and high sorbency polyester foam for excellent cleaning capacity and abrasion resistance. A textured option for applications that require improved particle removal is available.
Micronova™ NovaSorb™ String Mophead
Absorbent and lightweight, economically designed for single use in Controlled Environments.
Micronova™ SlimLine™ Mop Adapters
Plastic SlimLine adapter
Vileda Professional™ Extendable Universal Handle with Color Coding Rings
Provides an ergonomic feel and reliable telescoping performance. Vileda Professional-FHP Extendable Universal Handle with Color Coding Rings is part of the UltraSpeed Pro System and was developed to be simple to use.
Contec™ QuickConnect™ Mop Head Frames
Featuring a quick-connect button for easy attachment to all sizes and styles of Contec™ Quick Connect™ handles. Contec™ QuickConnect™ Mop Head Frames are specifically designed for use with the Contec™ family of mopping systems.
Contec™ VertiKlean™ Wall Washing Systems Sterile Mop Heads Available on GSA/VA Contract for Federal Government customers only.
Use with VertiKlean™ Wall Washing System which is designed for cleaning vertical surfaces. Contec™ VertiKlean™ Wall Washing Systems Sterile Mop Heads and Covers are comprised of laminated polyester fabric and high sorbency polyester foam for excellent cleaning capacity and abrasion resistance. A textured option for applications that require improved particle removal is available.
Vileda Professional™ Roll-O-Matic™ Self-Wringing Mops
Suitable for all controlled environments.
Foamtec PharmaMOP™ Cleanroom Mop Heads, Sterile
For cleaning and disinfection of rough surfaces and high soil areas
Micronova™ Tool/Mop Handles
Lightweight, plastic-coated fiberglass handles
Texwipe™ Replacement Covers for AlphaMop™ Cleanroom Mops
Suitable for requirements of Class 3-5 environments
Contec™ Edgeless™ Mop Handle with Frame
Integrated handle and frame is convenient and economical. Contec™ Edgeless™ Mop Handle with Frame are designed exclusively for Contec™ Edgeless™ Mop Heads.
Berkshire™ BCR™ Telescoping Aluminum Handle
BCR™ Telescoping aluminum handle extends from 37-71 in. /95-180 cm.
Contec™ VertiKlean™ Sox Mop Head Covers
Increases durability of VertiKlean mop when used on abrasive surfaces. Contec™ VertiKlean™ Sox Mop Head Covers are available in regular, medium and large to fit all VertiKlean mop heads.
Contec™ VertiKlean Sponges Available on GSA/VA Contract for Federal Government customers only.
Clean and wipe in laboratory settings with these absorbent, abrasion resistant sponges. Contec™ VertiKlean Sponges are made of absorbent polyester foam and abrasion resistant Polynit fabric. They are available in sterile and non-sterile varieties.
Micronova™ CurtainCleaner™ Handle
For use with CurtainCleaner Slipcovers
Micronova™ Accordion Mop Handle
Collapsible for use in isolators and containment hoods
Micronova™ SnapMop™ MegTex™ Irradiated Cleanroom Mop Heads
For cleaning rough/textured surfaces or removing heavy deposits
Micronova™ T-Mop™ Cleanroom Mops: Adapters
Low-profile, wide-surface mop handle
Micronova™ Mop Handles
Handle for Micronova mops
Micronova™ MicroMop™ and PolyGen™ Mop Handle
For use with Micronova string mops
Micronova™ NovaPoly™ Cleanroom Mop
Polyester-covered foam for critical applications and sterile support areas
Vileda Professional™ 10 in. Aluminum Extendable Roll-O-Matic™ Mop w/attached Refill
The best choice when seeking one mop to apply disinfectant, clean surfaces or absorb spills in all production and controlled environments!
Vileda Professional™ Evolution Telescopic Handle
Effective, ergonomic cleaning tool for window cleaning tasks. Vileda Professional Evolution Telescopic Handle is made of Aluminum.
  spinner