Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic Acid, 5% v/v (For AOAC Method 953.08), Fisher Chemical™
SDP

Catalog No. sa9050500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SA9050500 500 mL Natural Poly Bottle
SA90501 1 L Natural Poly Bottle
SA90504 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Alternative option to part number 400450
Catalog No. SA9050500
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Alternative option to part number 400450

Chemical Identifiers

CAS 7732-18-5 , 64-19-7
Molecular Formula C2H4O2
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

CAS Min % 94.0, 4.0
CAS Max % 96.0, 6.0
Quantity 500 mL
Packaging Natural Poly Bottle
Molecular Weight (g/mol) 60.05
Percent Purity 5% v/v
Grade Laboratory
Chemical Name or Material Acetic Acid, 5% v/v
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.