Learn More
Acetonitrile, LC-MS Ultra CHROMASOLV™, Honeywell Riedel-de Haën™
Shop All Honeywell Research Chemicals Products

Description
- For the most demanding LC-MS and UHPLC applications
- Tested for UHPLC-MS suitability
- Highest solvent purity for low detection limits
- Extremely low MS baseline noise
- Exemplary lot-to-lot reproducibility
- Low levels of trace metals
- Packaged in clear borosilicate glass bottles
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Density | 0.786g/mL at 25°C |
| Quantity | 2 L |
| Explosion Limit | 0.16 |
| Flash Point | 2°C (35.6°F) |
| Ignition Point | 973°F |
| Vapor Pressure | 72.8mmHg (20°C) |
| Linear Formula | CH3CN |
| UN Number | 1648 |
| Beilstein | 741857 |
| Solubility Information | Completely souble in water |
| Show More |
Safety and Handling
| Hazard Statement | P210-P280-P305 + P351 + P338 |
| Precautionary Statement | H225-H302 + H312 + H332-H319 |
| EINECSNumber | 200-835-2 |
| RTECSNumber | AL7700000 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.