Learn More
Description
Ahlstrom-Munksjö Grade 237 chromatography blotting paper is a medium-thick paper designed for general laboratory applications featuring medium-heavy solute loading.
- Grade: 237
- Thickness: 0.42 mm
- Absorption Speed: 150 mm/30 min
- Ideal Use: Serves as wicks in protein or nucleic acid blotting to ensure uniform flow of buffer through the gel to the transfer membrane in a blotting sandwich.
- Additional Applications: Suitable for electrophoresis, separation of inorganic compounds, Buchner funnels, and dye filtration.
- Performance Characteristics: High absorbency, high wet strength, and very high loading capacity.
- Whatman Equivalent: 3, 3CHR
Specifications
Specifications
| Length (Metric) | 10 cm |
| Width (Metric) | 7 cm |
| For Use With (Application) | Use as wicks in protein or nucleic acid blotting |
| Format | Sheet |
| Length (English) | 10 cm |
| Dimensions (LxW) | 7 x 10 cm |
| Thickness | 0.42 mm |
| Grade | 237 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.