Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer Solution, TISAB, Fisher Chemical™

Catalog No. p-212983 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
4 L
20 L
Packaging:
Poly Bottle
PolyPac™
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SB175-4 4 L Poly Bottle
SB17520 20 L PolyPac™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SB175-4
Add to Cart

missing translation for 'validMainframeMsg'

For Analysis of the Determination of Fluoride Ion Concentration by Fluoride Ion Electrode

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Buffer Solution, TISAB
Name Note Total Ionic Strength Adjustment Buffer
CAS 7647-14-5, 127-09-3, 482-54-2, 9003-39-8, 52-51-7
Color Undesignated
pH 5.5
Physical Form Liquid
Quantity 4 L
Identification Pass Test
MDL Number MFCD00146206
Packaging Poly Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.