Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Globally Harmonized System Warning Pictogram Poster
SCI_ED

Catalog No. s08085 Shop All Fisherbrand Products
Click to view available options
Grade:
Middle School and Up
This item is not returnable. View return policy

Catalog No. S08085

Only null left
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Explains the nine Physical, Health and Environmental Hazard pictograms as part of the GHS system of classification/labeling of chemicals.

  • Features the nine pictograms of the GHS system of classification/labeling of chemicals
  • Heavy glossy paper stock
  • Size: 24x36 in.

Specifications

Product Type GHS Pictogram Poster
Length (English) 24 in.
Width (English) 36 in.
Width (Metric) 36 in.
Grade Middle School and Up
Material Glossy Stock Paper
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.