Learn More
Description
- For gas chromatography (GC)
- Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
- Non-volatile matter and water content are specified for each solvent
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Density | 0.659g/mL at 25°C |
| Quantity | 4 x 4 L |
| Explosion Limit | 0.077 |
| Flash Point | −26°C (−14.8°F) |
| Ignition Point | 453°F |
| Linear Formula | CH3(CH2)4CH3 |
| Vapor Pressure | 5.2 psi (37.7°C), 256 mmHg (37.7°C), 132 mmHg (20°C) |
| UN Number | UN1208 |
| Beilstein | 1730733 |
| Formula Weight | 86.18g/mol |
| Show More |
Safety and Handling
| Hazard Statement | P201-P210-P273-P301 + P310-P308 + P313-P331 |
| RTECSNumber | MN9275000 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
