Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodine (Resublimed Crystals/USP), Fisher Chemical™
GSA_VA

Catalog No. p-32011 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
I35-100 100 g Glass Bottle
I35-500 500 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. I35-100
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7553-56-2
Molecular Formula I2
Molecular Weight (g/mol) 253.81
MDL Number MFCD00011355 MFCD00164163
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
Synonym iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
PubChem CID 807
ChEBI CHEBI:17606
IUPAC Name diiodine
SMILES II

Specifications

Boiling Point 184°C
Melting Point 113°C
Color Black
Physical Form Solid
Quantity 100 g
Assay Percent Range 99.8 to 100.5%
UN Number 1759
Formula Weight 253.81
Percent Purity 99.8 to 100.5%
Grade USP
Identification Pass test
Packaging Glass Bottle
Vapor Pressure 0.3mmHg at 20°C
Density 4.93g/cm³; Vapor Density: 8.8g/mol
Assay 99.8-100.5%
Chemical Name or Material Iodine
Show More Show Less
Safety handling Check Alert storage code Red
DOTInformation DOT Class 8, : Corrosive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.