Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
METTLER TOLEDO™ SevenCompact™ S220 pH Benchtop Meter
Shop All METTLER TOLEDO Products
Click to view available options
Model:
Bio/Life Science Option
Food (Solid/Viscous) Option
General Applications Option
Description
- pH, Ion Concentration, and Redox/ORP benchtop meter that can display results in pH, mV, and Rel mV
- Durable and simple to operate with two buttons: 'Read' and 'Cal'
- IP54 Protection
- Comes with electrode arm that moves straight up and down, bringing the electrode into position where it works best for your sample
- Compatible with a wide range of electrodes for any application
- Compatible with ISM electrodes that capture and save calibration data, which is then automatically recognized by the meter once connected
- Meter connects to electrodes via BNC; S7 to BNC cables available
- Results can be captured by a printer with a time and date stamp
- Optional installation and qualification (IQ/OQ) package available through METTLER TOLEDO Service
Specifications
Specifications
| Accuracy (mV) | ±0.2 mV |
| Accuracy (pH) | ±0.002 |
| Description | S220 Kit - Bio & Life Science Measurements |
| Display Type | TFT |
| Range (mV) | -2000 to 2000 mV |
| Range (pH) | -2 to 20 |
| Resolution (pH) | 0.001/0.01/0.1 |
| Temperature Range (Metric) | -30.0°C to +130°C (MTC), -5.0°C to +130.0°C (ATC) |
| For Use With (Application) | Bio & Life Science |
| Includes | S220 pH meter, InLab Routine Pro-ISM electrode, Multipin-BNC/RCA electrode cable, electrode holder, 2 buffer sachets for pH 4.01, 7, 9.21 and 10.01, pH Measurement Guide, protective cover, test certificate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction