Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Reichert™ Darkfield Quebec™ Colony Counters

Catalog No. p-139509 Shop All Reichert, Inc Products
Change view
Click to view available options
Model:
Electronic
Standard
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
RE-3325 Standard
07-908-7 Electronic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. RE-3325 Supplier Reichert™ Supplier No. 13332500
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Includes

Both models are supplied with white-ruled Wolffheugel guide plate, three-wire cord and 1.5X lens. Electronic Register model also includes probe and ground.

Count bacterial colonies quickly and accurately

Exclusive darkfield design provides even, glare-free illumination. Contrasted against darkfield background, colonies are bright—readily distinguished from other structures in agar. Each probing action into the colony completes a circuit and is automatically recorded (Electronic Register model only).

Key Features (both models):

  • Highly polished, annular reflector spreads light from 40-watt bulb uniformly over entire culture plate
  • Operating switch conveniently mounted on front of instrument case
  • Hinged door provides easy access to change light bulb—just remove two screws
  • Container holder adapts for centering round dishes up to 100mm diameter or square culture plates up to 100 × 100mm
  • Adjustable focusing rod allows 1.5X lens to be raised or lowered for magnified viewing
  • Lens rotates 360° for ready access to culture plates
  • Reset bar provides immediate zeroing of five-digit counter
  • Built-in tilt leg may be mounted in front or rear; allows convenient tilt angle or may be locked flat to instrument base

Electronic register model

  • Operated by needle-point probe with magnetic ground
  • When ground and counting probes both touch conductive agar in dish or plate, an electrical circuit is completed through the integral amplifier to activate the digital counter

Certifications

UL Listed

Specifications

Product Type Darkfield Colony Counter
Model Standard
Height (English) 10 in.
Height (Metric) 25cm
Length (English) 10.5 in.
Length (Metric) 27cm
Width (English) 9.25 in.
Width (Metric) 23cm
Weight (English) 12 lb.
Weight (Metric) 5500g
Frequency 60Hz
Voltage 115V
Certifications/Compliance UL
For Use With (Application) Count bacterial colonies quickly and accurately
Includes USA cord, Wolffheugel counting plate, three-wire cord and 1.5X lens
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.