Learn More
Thermo Scientific™ SMART Digest™ Trypsin Kits
Description
Obtain fast, simple, and highly reproducible trypsin protein digestion for peptide characterization and quantitation in biopharmaceutical protein research.
Obtain high quality analytical results from protein digests using Thermo Scientific™ SMART Digest™ Kits. A significant advance in sample preparation for biopharmaceutical protein research, the kits provide fast and simple protein digestion with high reproducibility, high sensitivity, and high levels of data quality in a format that's compatible with automation. The SMART Digest Kits are a significant improvement over current in-solution protein digestion technologies, which are not reproducible, have poor sensitivity, and employ protracted methodologies not amenable to automation.
SMART Digest kits enable you to characterize and quantitate proteins faster, easier, and quicker than ever before. This version of the kit uses trypsin, which cleaves peptides on the C-terminal side of lysine and arginine amino acid residues.
Easy and simple- Significantly increased speed of sample processing
- Simple process
- Easily automate entire protocol for high-throughput processing
- Increased reproducibility over existing protocols
- Higher sensitivity
- Higher data confidence
Typically it takes less than 60 minutes (dependent on sample complexity) to achieve full digestion, whereas an in-solution digest can take up to 24 hours.
Cleaner SamplesReduction in the number of chemicals used and the immobilized trypsin design helps mitigate against chemically induced post translational modifications (PTMs) and autolysis. This results in results which are easier to interpret.
Couple with SOLAμ SPE Plates for DesaltingWhen used with the SMART Digest kits for desalting, Thermo Scientific™ SOLAμ™ SPE solid phase extraction plates provide a fast, easy-to-use, robust, and easily automatable sample preparation solution for analysis of biomolecules.
Specifications
Specifications
| Type | Trypsin Kit |
| Description | SMART Digest Trypsin Kit, with Collection plate |
| Format | Resin in Pre-filled PCR Tubes |
| Enzyme | Trypsin |
| Cleavage | C-terminal side of lysine and arginine amino acids |
| Capture | None |
| Product Line | SMART Digest |
| For Use With (Application) | Peptide Mapping |
| Includes | Resin, Collection Plate |
Frequently Asked Questions (FAQs)
• CHAPS – results in ~ 30% reduction in digest efficiency.
• OGS – no reduction in digestion efficiency.
• TWEEN – no reduction in digestion efficiency.
• RIPA – results in ~ 20% reduction in digestion efficiency. The addition of RIPA, for ribonuclease A digestion, results in a concentration-dependent effect, where initial enzyme inhibition is overcome by improved substrate solubilization at higher concentrations only.
The SMART Digest Trypsin Kit is not affected by concentrations of up to 0.5 M of urea. If the concentration is higher than this, we recommend that the sample is diluted to less than 0.5 M of urea prior to beginning digestion using the SMART Digest Trypsin Kit.
The composition of the SMART Digest Trypsin Kit buffer is as follows:
Chemical Name (CAS No., EINECS No.): Kit Component Quantity, Weight %
• Water (7732-18-5, 231-791-2): 2, 50-95%
• Glycerol (56-81-5, 200-289-5): 2, < 20%
• Tris Base (77-86-1, 201-064-4): 2, < 10%
• Tris-HCI (1185-53-1, 214-684-5): 2, < 10%
• Calcium Chloride (10043-52-4, 233-140-8): 2, < 10%
• Sodium Azide (26628-22-8, 247-852-1): 2, < 0.1%
The SMART Digest Trypsin Kit contains an excess of enzyme capable of digesting between 200 pg and 3.5 mg of protein. As most samples will fall in this range, it is not necessary to routinely quantitate protein concentration prior to digestion.
The reason urea is needed as a first step in an in-solution protocol is to disrupt the sample and partially unfold the proteins. The proteins need to be partially unfolded so that the trypsin enzyme can have better access to the internal amino acid chain, not just the surface of the protein of interest. The reason the SMART Digest Trypsin kit does not need urea is that it uses heat to unfold the protein.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.