This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ARF5 SIRNA Assays ›
Gene Symbol:
ARF5
Gene Name:
ADP ribosylation factor 5
Gene Aliases:
-
Chromosome Location:
Chr.7: 127588352 - 127591705 on Build GRCh38
UniGene ID:
Hs.653202
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s1562
Validated

Purification
Size
Availability Inventoried
Catalog # 4390824
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ARF5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001662.3 NP_001653.1 4 421
GenBank mRNA AF493884.1 - 4 274
AK297026.1 - 4 326
AK307297.1 - 3 866
AK308548.1 - 3 1074
AK312107.1 - 4 326
BC003043.1 - 4 311 2822973
BC033104.1 - 4 368 4862014
BT007087.1 - 4 274
CN259625.1 - 4 438
M57567.1 - 4 328

Back To Top

More Information


Matching Taqman Gene Expression Assay:


Hs00155845_m1

Panther Classification:

Molecular Function -

G-protein

Gene Ontology Categories:

Function(s) Process(es)

retrograde vesicle-mediated transport, Golgi to ER
small GTPase mediated signal transduction
protein transport
GTPase activity
protein binding
GTP binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-pxtp8:80/100.66.75.48:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline